Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35176672-35176897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD44
Entrez Gene ID
960
Gene Name
CD44 molecule (Indian blood group)
Gene Aliases
CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1
UniGene
Hs.502328
Gene Chromosome Location
Chr.11: 35138870-35232402 on Build GRCh38
Transcripts
NM_000610.3 NM_001001389.1 NM_001001390.1 NM_001001391.1 NM_001001392.1 NM_001202555.1 NM_001202556.1 NM_001202557.1 XM_005253231.2 XM_005253232.2 XM_005253233.2 XM_005253234.2 XM_005253235.2 XM_005253238.2 XM_005253239.2 XM_005253240.2 XM_006718388.1 XM_006718390.3 XM_011520482.1 XM_011520483.1 XM_011520484.1 XM_011520485.1 XM_011520486.1 XM_011520487.2 XM_011520488.1 XM_011520489.2 XM_017018583.1 XM_017018584.1 XM_017018585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3809156 c.199G>T p.E67* Chr.11: 35176706-35176706 on Build GRCh38
6241822 c.212G>A p.S71N Chr.11: 35176719-35176719 on Build GRCh38
4187719 c.233G>A p.R78K Chr.11: 35176740-35176740 on Build GRCh38
1492531 c.221T>A p.F74Y Chr.11: 35176728-35176728 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760222232 Chr.11: 35176698-35176698 on Build GRCh38
rs780409682 Chr.11: 35176730-35176730 on Build GRCh38
rs967250097 Chr.11: 35176693-35176693 on Build GRCh38
rs568705475 Chr.11: 35176857-35176857 on Build GRCh38
rs748214228 Chr.11: 35176756-35176756 on Build GRCh38
rs756890802 Chr.11: 35176723-35176723 on Build GRCh38
rs778704529 Chr.11: 35176743-35176743 on Build GRCh38
rs753441551 Chr.11: 35176708-35176708 on Build GRCh38
rs375417895 Chr.11: 35176777-35176777 on Build GRCh38
rs138821226 Chr.11: 35176697-35176697 on Build GRCh38
rs1055561529 Chr.11: 35176798-35176798 on Build GRCh38
rs755323887 Chr.11: 35176858-35176858 on Build GRCh38
rs763715662 Chr.11: 35176706-35176706 on Build GRCh38
rs369701383 Chr.11: 35176784-35176784 on Build GRCh38
rs916654275 Chr.11: 35176818-35176818 on Build GRCh38
rs548473416 Chr.11: 35176755-35176755 on Build GRCh38
rs74594564 Chr.11: 35176792-35176792 on Build GRCh38
rs1033752531 Chr.11: 35176817-35176817 on Build GRCh38
rs749457039 Chr.11: 35176758-35176758 on Build GRCh38
rs758352011 Chr.11: 35176745-35176745 on Build GRCh38
rs928571731 Chr.11: 35176721-35176721 on Build GRCh38
rs750242872 Chr.11: 35176744-35176744 on Build GRCh38
rs1006831435 Chr.11: 35176807-35176807 on Build GRCh38
rs768073670 Chr.11: 35176696-35176696 on Build GRCh38
rs374806320 Chr.11: 35176757-35176757 on Build GRCh38

View Full Product Details