Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83273447-83273700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ2
Entrez Gene ID
27235
Gene Name
coenzyme Q2, polyprenyltransferase
Gene Aliases
CL640, COQ10D1, MSA1, PHB:PPT
UniGene
Hs.144304
Gene Chromosome Location
Chr.4: 83138568-83285129 on Build GRCh38
Transcripts
NM_015697.7 XM_011531855.2 XM_011531857.2 XM_011531859.2 XM_011531860.2 XM_011531862.2 XM_011531863.2 XM_011531866.2 XM_011531867.2 XM_017008031.1 XM_017008032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
222035 c.637C>T p.L213F Chr.4: 83273551-83273551 on Build GRCh38
1633817 c.661C>G p.L221V Chr.4: 83273527-83273527 on Build GRCh38
1633816 c.665G>T p.G222V Chr.4: 83273523-83273523 on Build GRCh38
3606591 c.627C>T p.S209S Chr.4: 83273561-83273561 on Build GRCh38
4572192 c.671T>A p.L224H Chr.4: 83273517-83273517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376948148 Chr.4: 83273588-83273588 on Build GRCh38
rs768637231 Chr.4: 83273662-83273662 on Build GRCh38
rs188476963 Chr.4: 83273537-83273537 on Build GRCh38
rs770226811 Chr.4: 83273643-83273643 on Build GRCh38
rs774470336 Chr.4: 83273585-83273585 on Build GRCh38
rs778590200 Chr.4: 83273615-83273622 on Build GRCh38
rs764312091 Chr.4: 83273584-83273584 on Build GRCh38
rs200791648 Chr.4: 83273544-83273544 on Build GRCh38
rs375674679 Chr.4: 83273540-83273540 on Build GRCh38
rs144913206 Chr.4: 83273614-83273614 on Build GRCh38
rs754448712 Chr.4: 83273606-83273606 on Build GRCh38
rs775863297 Chr.4: 83273655-83273655 on Build GRCh38
rs746359697 Chr.4: 83273640-83273640 on Build GRCh38
rs372949213 Chr.4: 83273587-83273587 on Build GRCh38
rs769049644 Chr.4: 83273634-83273635 on Build GRCh38
rs768409758 Chr.4: 83273521-83273521 on Build GRCh38
rs567972522 Chr.4: 83273560-83273560 on Build GRCh38
rs747330347 Chr.4: 83273528-83273528 on Build GRCh38
rs757009353 Chr.4: 83273496-83273496 on Build GRCh38
rs121918231 Chr.4: 83273598-83273598 on Build GRCh38
rs778586293 Chr.4: 83273626-83273626 on Build GRCh38
rs765970358 Chr.4: 83273599-83273599 on Build GRCh38
rs1003416138 Chr.4: 83273673-83273673 on Build GRCh38
rs376446272 Chr.4: 83273660-83273660 on Build GRCh38
rs747345294 Chr.4: 83273632-83273633 on Build GRCh38
rs372421130 Chr.4: 83273485-83273485 on Build GRCh38
rs750411784 Chr.4: 83273504-83273504 on Build GRCh38
rs747777883 Chr.4: 83273637-83273637 on Build GRCh38
rs778440756 Chr.4: 83273526-83273526 on Build GRCh38
rs748884504 Chr.4: 83273511-83273511 on Build GRCh38
rs779747173 Chr.4: 83273507-83273507 on Build GRCh38
rs774606974 Chr.4: 83273666-83273666 on Build GRCh38
rs376202439 Chr.4: 83273582-83273582 on Build GRCh38
rs775606443 Chr.4: 83273572-83273572 on Build GRCh38
rs369421547 Chr.4: 83273484-83273484 on Build GRCh38
rs757596156 Chr.4: 83273638-83273638 on Build GRCh38
rs762958081 Chr.4: 83273573-83273573 on Build GRCh38
rs893413982 Chr.4: 83273613-83273613 on Build GRCh38
rs539904082 Chr.4: 83273639-83273639 on Build GRCh38
rs746905594 Chr.4: 83273549-83273549 on Build GRCh38
rs121918232 Chr.4: 83273505-83273505 on Build GRCh38
rs899664560 Chr.4: 83273550-83273550 on Build GRCh38
rs150295921 Chr.4: 83273601-83273601 on Build GRCh38

View Full Product Details