Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57744814-57745039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD18
Entrez Gene ID
643376
Gene Name
BTB domain containing 18
Gene Aliases
-
UniGene
Hs.606355
Gene Chromosome Location
Chr.11: 57743514-57753176 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7190188 c.1417C>A p.L473M Chr.11: 57744856-57744856 on Build GRCh38
5967918 c.1282G>A p.D428N Chr.11: 57744991-57744991 on Build GRCh38
4922181 c.1307C>A p.S436Y Chr.11: 57744966-57744966 on Build GRCh38
1185035 c.1434T>A p.C478* Chr.11: 57744839-57744839 on Build GRCh38
6007728 c.1343C>A p.S448Y Chr.11: 57744930-57744930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010354997 Chr.11: 57744958-57744958 on Build GRCh38
rs955459777 Chr.11: 57744982-57744982 on Build GRCh38
rs1000483658 Chr.11: 57744895-57744895 on Build GRCh38
rs903491338 Chr.11: 57744887-57744887 on Build GRCh38
rs748317389 Chr.11: 57745004-57745004 on Build GRCh38
rs781759530 Chr.11: 57744922-57744922 on Build GRCh38
rs774642543 Chr.11: 57744955-57744955 on Build GRCh38
rs955156676 Chr.11: 57744975-57744975 on Build GRCh38
rs749751437 Chr.11: 57744865-57744865 on Build GRCh38
rs769269326 Chr.11: 57744932-57744932 on Build GRCh38
rs769328429 Chr.11: 57744894-57744894 on Build GRCh38
rs1029752492 Chr.11: 57744968-57744968 on Build GRCh38
rs578128448 Chr.11: 57744872-57744872 on Build GRCh38
rs756881265 Chr.11: 57744843-57744843 on Build GRCh38
rs753698203 Chr.11: 57744860-57744874 on Build GRCh38
rs780590058 Chr.11: 57744859-57744859 on Build GRCh38
rs780706565 Chr.11: 57744854-57744854 on Build GRCh38
rs781430128 Chr.11: 57745008-57745008 on Build GRCh38
rs747341415 Chr.11: 57744892-57744892 on Build GRCh38

View Full Product Details