Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92379616-92379879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 XM_011518689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6116636 c.532G>C p.E178Q Chr.9: 92379827-92379827 on Build GRCh38
1463576 c.513delT p.F173fs*46 Chr.9: 92379808-92379808 on Build GRCh38
6626687 c.560T>G p.L187R Chr.9: 92379855-92379855 on Build GRCh38
3382602 c.505A>C p.S169R Chr.9: 92379800-92379800 on Build GRCh38
6027644 c.524A>T p.E175V Chr.9: 92379819-92379819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751178579 Chr.9: 92379747-92379747 on Build GRCh38
rs772146071 Chr.9: 92379834-92379834 on Build GRCh38
rs750046294 Chr.9: 92379729-92379729 on Build GRCh38
rs530316962 Chr.9: 92379744-92379744 on Build GRCh38
rs376115969 Chr.9: 92379773-92379773 on Build GRCh38
rs764974855 Chr.9: 92379720-92379720 on Build GRCh38
rs74959855 Chr.9: 92379839-92379839 on Build GRCh38
rs531329924 Chr.9: 92379779-92379779 on Build GRCh38
rs1051627997 Chr.9: 92379642-92379642 on Build GRCh38
rs745890139 Chr.9: 92379833-92379833 on Build GRCh38
rs909176139 Chr.9: 92379830-92379830 on Build GRCh38
rs772923884 Chr.9: 92379847-92379847 on Build GRCh38
rs377479211 Chr.9: 92379719-92379719 on Build GRCh38
rs200177607 Chr.9: 92379792-92379792 on Build GRCh38
rs778019537 Chr.9: 92379787-92379787 on Build GRCh38
rs41278665 Chr.9: 92379730-92379730 on Build GRCh38
rs898452267 Chr.9: 92379708-92379708 on Build GRCh38
rs760602540 Chr.9: 92379712-92379712 on Build GRCh38
rs571166588 Chr.9: 92379805-92379805 on Build GRCh38
rs774476954 Chr.9: 92379818-92379818 on Build GRCh38
rs750494120 Chr.9: 92379808-92379808 on Build GRCh38
rs202174613 Chr.9: 92379799-92379799 on Build GRCh38
rs766049231 Chr.9: 92379843-92379843 on Build GRCh38
rs999313294 Chr.9: 92379679-92379679 on Build GRCh38
rs563515121 Chr.9: 92379759-92379759 on Build GRCh38
rs769828488 Chr.9: 92379714-92379714 on Build GRCh38
rs776564796 Chr.9: 92379718-92379718 on Build GRCh38
rs902247657 Chr.9: 92379678-92379678 on Build GRCh38
rs983950200 Chr.9: 92379804-92379804 on Build GRCh38
rs767597478 Chr.9: 92379725-92379727 on Build GRCh38
rs112987350 Chr.9: 92379840-92379840 on Build GRCh38
rs374571991 Chr.9: 92379802-92379802 on Build GRCh38
rs370633429 Chr.9: 92379841-92379841 on Build GRCh38
rs143792971 Chr.9: 92379798-92379798 on Build GRCh38
rs371577890 Chr.9: 92379797-92379797 on Build GRCh38
rs183257910 Chr.9: 92379749-92379749 on Build GRCh38
rs201096846 Chr.9: 92379742-92379742 on Build GRCh38

View Full Product Details