Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11421532-11421793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC151
Entrez Gene ID
115948
Gene Name
coiled-coil domain containing 151
Gene Aliases
CILD30
UniGene
Hs.124010
Gene Chromosome Location
Chr.19: 11420605-11435782 on Build GRCh38
Transcripts
NM_001302453.1 NM_001302454.1 NM_145045.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1247477 c.1507G>A p.V503M Chr.19: 11421760-11421760 on Build GRCh38
2814475 c.1505T>G p.L502R Chr.19: 11421762-11421762 on Build GRCh38
5741443 c.1493A>C p.N498T Chr.19: 11421774-11421774 on Build GRCh38
4389417 c.1587C>T p.R529R Chr.19: 11421680-11421680 on Build GRCh38
6274716 c.1590+4C>A p.? Chr.19: 11421673-11421673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772230508 Chr.19: 11421720-11421720 on Build GRCh38
rs77133587 Chr.19: 11421766-11421766 on Build GRCh38
rs1047577481 Chr.19: 11421554-11421554 on Build GRCh38
rs747116785 Chr.19: 11421730-11421730 on Build GRCh38
rs764166460 Chr.19: 11421654-11421654 on Build GRCh38
rs779597814 Chr.19: 11421667-11421667 on Build GRCh38
rs756578727 Chr.19: 11421676-11421676 on Build GRCh38
rs955753708 Chr.19: 11421747-11421747 on Build GRCh38
rs779209740 Chr.19: 11421710-11421710 on Build GRCh38
rs1029818682 Chr.19: 11421733-11421733 on Build GRCh38
rs975331253 Chr.19: 11421620-11421620 on Build GRCh38
rs772186012 Chr.19: 11421642-11421642 on Build GRCh38
rs765069377 Chr.19: 11421759-11421759 on Build GRCh38
rs768657788 Chr.19: 11421732-11421732 on Build GRCh38
rs751016897 Chr.19: 11421770-11421770 on Build GRCh38
rs775489013 Chr.19: 11421645-11421645 on Build GRCh38
rs1010583935 Chr.19: 11421649-11421649 on Build GRCh38
rs761910295 Chr.19: 11421752-11421752 on Build GRCh38
rs558928167 Chr.19: 11421661-11421661 on Build GRCh38
rs368706462 Chr.19: 11421760-11421760 on Build GRCh38
rs759465416 Chr.19: 11421637-11421637 on Build GRCh38
rs761714197 Chr.19: 11421657-11421657 on Build GRCh38
rs201300238 Chr.19: 11421630-11421630 on Build GRCh38
rs778270617 Chr.19: 11421682-11421682 on Build GRCh38
rs564009092 Chr.19: 11421608-11421608 on Build GRCh38
rs749593551 Chr.19: 11421691-11421691 on Build GRCh38
rs531115348 Chr.19: 11421648-11421648 on Build GRCh38
rs774643840 Chr.19: 11421632-11421632 on Build GRCh38
rs753638961 Chr.19: 11421655-11421655 on Build GRCh38
rs776715104 Chr.19: 11421736-11421736 on Build GRCh38
rs151110715 Chr.19: 11421715-11421715 on Build GRCh38
rs941051010 Chr.19: 11421726-11421726 on Build GRCh38
rs367922109 Chr.19: 11421666-11421666 on Build GRCh38
rs1022093761 Chr.19: 11421599-11421599 on Build GRCh38
rs375632703 Chr.19: 11421684-11421684 on Build GRCh38
rs751113796 Chr.19: 11421669-11421669 on Build GRCh38
rs775607293 Chr.19: 11421725-11421725 on Build GRCh38
rs750087328 Chr.19: 11421660-11421660 on Build GRCh38
rs908248464 Chr.19: 11421709-11421709 on Build GRCh38
rs974238862 Chr.19: 11421742-11421742 on Build GRCh38
rs371107994 Chr.19: 11421672-11421672 on Build GRCh38
rs757642892 Chr.19: 11421694-11421694 on Build GRCh38
rs764039877 Chr.19: 11421765-11421765 on Build GRCh38
rs762638578 Chr.19: 11421761-11421761 on Build GRCh38
rs764731001 Chr.19: 11421659-11421659 on Build GRCh38
rs906414976 Chr.19: 11421578-11421578 on Build GRCh38

View Full Product Details