Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203217668-203217941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIT1
Entrez Gene ID
1118
Gene Name
chitinase 1
Gene Aliases
CHI3, CHIT, CHITD
UniGene
Hs.201688
Gene Chromosome Location
Chr.1: 203216079-203229732 on Build GRCh38
Transcripts
NM_001256125.1 NM_001270509.1 NM_003465.2 NR_045784.1 NR_045785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628401 c.1099G>A p.G367S Chr.1: 203217796-203217796 on Build GRCh38
4611382 c.1062_1063ins20 p.W358fs*1 Chr.1: 203217832-203217833 on Build GRCh38
5827703 c.1046G>A p.W349* Chr.1: 203217822-203217822 on Build GRCh38
6369654 c.1031G>T p.G344V Chr.1: 203217837-203217837 on Build GRCh38
1745478 c.1072_1073ins24 p.V357_W358ins* Chr.1: 203217822-203217823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748129910 Chr.1: 203217912-203217912 on Build GRCh38
rs748935586 Chr.1: 203217743-203217744 on Build GRCh38
rs764257734 Chr.1: 203217695-203217695 on Build GRCh38
rs537102838 Chr.1: 203217856-203217856 on Build GRCh38
rs757044414 Chr.1: 203217898-203217898 on Build GRCh38
rs781266995 Chr.1: 203217847-203217848 on Build GRCh38
rs375923575 Chr.1: 203217879-203217879 on Build GRCh38
rs762064575 Chr.1: 203217829-203217829 on Build GRCh38
rs149987600 Chr.1: 203217770-203217770 on Build GRCh38
rs752734289 Chr.1: 203217898-203217898 on Build GRCh38
rs756629559 Chr.1: 203217715-203217715 on Build GRCh38
rs766945133 Chr.1: 203217775-203217775 on Build GRCh38
rs770731880 Chr.1: 203217805-203217805 on Build GRCh38
rs2297947 Chr.1: 203217735-203217735 on Build GRCh38
rs779679941 Chr.1: 203217729-203217729 on Build GRCh38
rs142332008 Chr.1: 203217807-203217807 on Build GRCh38
rs374021722 Chr.1: 203217756-203217756 on Build GRCh38
rs150192398 Chr.1: 203217822-203217823 on Build GRCh38
rs147080247 Chr.1: 203217836-203217836 on Build GRCh38
rs984967613 Chr.1: 203217699-203217699 on Build GRCh38
rs577366461 Chr.1: 203217885-203217885 on Build GRCh38
rs767840128 Chr.1: 203217713-203217713 on Build GRCh38
rs3831317 Chr.1: 203217846-203217847 on Build GRCh38
rs760891565 Chr.1: 203217849-203217849 on Build GRCh38
rs771290544 Chr.1: 203217766-203217767 on Build GRCh38
rs985439446 Chr.1: 203217892-203217892 on Build GRCh38
rs199600055 Chr.1: 203217798-203217798 on Build GRCh38
rs982000660 Chr.1: 203217722-203217722 on Build GRCh38
rs752379824 Chr.1: 203217796-203217796 on Build GRCh38
rs137954453 Chr.1: 203217862-203217862 on Build GRCh38
rs779206133 Chr.1: 203217778-203217778 on Build GRCh38
rs201561248 Chr.1: 203217719-203217719 on Build GRCh38
rs757953527 Chr.1: 203217888-203217888 on Build GRCh38
rs764818130 Chr.1: 203217871-203217871 on Build GRCh38
rs150852382 Chr.1: 203217751-203217751 on Build GRCh38
rs767902609 Chr.1: 203217772-203217772 on Build GRCh38
rs386369359 Chr.1: 203217823-203217824 on Build GRCh38
rs1023095364 Chr.1: 203217704-203217704 on Build GRCh38
rs993853378 Chr.1: 203217789-203217789 on Build GRCh38
rs747662521 Chr.1: 203217726-203217727 on Build GRCh38
rs780072786 Chr.1: 203217905-203217905 on Build GRCh38
rs1022174330 Chr.1: 203217758-203217758 on Build GRCh38
rs778631916 Chr.1: 203217785-203217785 on Build GRCh38
rs1048695651 Chr.1: 203217710-203217710 on Build GRCh38
rs200696379 Chr.1: 203217894-203217894 on Build GRCh38
rs772208489 Chr.1: 203217918-203217918 on Build GRCh38
rs750793447 Chr.1: 203217774-203217774 on Build GRCh38
rs139939985 Chr.1: 203217750-203217750 on Build GRCh38
rs758032638 Chr.1: 203217797-203217797 on Build GRCh38
rs367900575 Chr.1: 203217769-203217769 on Build GRCh38
rs140228721 Chr.1: 203217851-203217851 on Build GRCh38
rs928768086 Chr.1: 203217727-203217727 on Build GRCh38
rs745787601 Chr.1: 203217899-203217899 on Build GRCh38
rs772938001 Chr.1: 203217755-203217755 on Build GRCh38
rs750932103 Chr.1: 203217714-203217714 on Build GRCh38
rs201320385 Chr.1: 203217822-203217822 on Build GRCh38
rs769763032 Chr.1: 203217903-203217903 on Build GRCh38
rs866287162 Chr.1: 203217813-203217813 on Build GRCh38
rs968745998 Chr.1: 203217882-203217882 on Build GRCh38
rs754057913 Chr.1: 203217836-203217837 on Build GRCh38
rs896914974 Chr.1: 203217786-203217786 on Build GRCh38
rs754408295 Chr.1: 203217777-203217777 on Build GRCh38
rs200625369 Chr.1: 203217740-203217740 on Build GRCh38
rs772902435 Chr.1: 203217741-203217742 on Build GRCh38
rs41304073 Chr.1: 203217897-203217897 on Build GRCh38
rs371660597 Chr.1: 203217828-203217828 on Build GRCh38
rs371800652 Chr.1: 203217703-203217703 on Build GRCh38
rs770891150 Chr.1: 203217767-203217767 on Build GRCh38
rs911031003 Chr.1: 203217844-203217844 on Build GRCh38
rs888078038 Chr.1: 203217814-203217814 on Build GRCh38
rs143439055 Chr.1: 203217737-203217740 on Build GRCh38
rs748770137 Chr.1: 203217730-203217730 on Build GRCh38
rs745875622 Chr.1: 203217809-203217809 on Build GRCh38
rs201682373 Chr.1: 203217819-203217819 on Build GRCh38
rs9943208 Chr.1: 203217835-203217835 on Build GRCh38
rs368445196 Chr.1: 203217697-203217697 on Build GRCh38
rs983971974 Chr.1: 203217840-203217840 on Build GRCh38
rs997457411 Chr.1: 203217747-203217747 on Build GRCh38
rs747908836 Chr.1: 203217848-203217848 on Build GRCh38
rs763127624 Chr.1: 203217689-203217689 on Build GRCh38
rs760729445 Chr.1: 203217835-203217836 on Build GRCh38
rs771895868 Chr.1: 203217753-203217753 on Build GRCh38
rs868208340 Chr.1: 203217845-203217846 on Build GRCh38

View Full Product Details