Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 54255515-54255776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACYP2
Entrez Gene ID
98
Gene Name
acylphosphatase 2
Gene Aliases
ACYM, ACYP
UniGene
Hs.516173
Gene Chromosome Location
Chr.2: 53970838-54305300 on Build GRCh38
Transcripts
NM_001320586.1 NM_001320587.1 NM_001320588.1 NM_001320589.1 NM_138448.3 XM_017005411.1 XM_017005412.1 XM_017005413.1 NM_001003937.2

Gene Symbol
TSPYL6
Entrez Gene ID
388951
Gene Name
TSPY like 6
Gene Aliases
-
UniGene
Hs.620508
Gene Chromosome Location
Chr.2: 54253178-54256272 on Build GRCh38
Transcripts
NM_001320586.1 NM_001320587.1 NM_001320588.1 NM_001320589.1 NM_138448.3 XM_017005411.1 XM_017005412.1 XM_017005413.1 NM_001003937.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5658395 c.463T>C p.C155R Chr.2: 54255689-54255689 on Build GRCh38
1268812 c.504C>T p.G168G Chr.2: 54255648-54255648 on Build GRCh38
1230719 c.580G>A p.G194R Chr.2: 54255572-54255572 on Build GRCh38
6884716 c.459G>A p.E153E Chr.2: 54255693-54255693 on Build GRCh38
3052376 c.440C>T p.A147V Chr.2: 54255712-54255712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768887118 Chr.2: 54255696-54255696 on Build GRCh38
rs79371652 Chr.2: 54255579-54255579 on Build GRCh38
rs376982052 Chr.2: 54255546-54255546 on Build GRCh38
rs766116667 Chr.2: 54255568-54255585 on Build GRCh38
rs757430041 Chr.2: 54255567-54255578 on Build GRCh38
rs752676197 Chr.2: 54255607-54255607 on Build GRCh38
rs754452381 Chr.2: 54255600-54255600 on Build GRCh38
rs751318047 Chr.2: 54255580-54255585 on Build GRCh38
rs768660841 Chr.2: 54255655-54255655 on Build GRCh38
rs764749605 Chr.2: 54255720-54255720 on Build GRCh38
rs764712537 Chr.2: 54255602-54255602 on Build GRCh38
rs775497090 Chr.2: 54255637-54255637 on Build GRCh38
rs187843016 Chr.2: 54255744-54255744 on Build GRCh38
rs375734975 Chr.2: 54255613-54255613 on Build GRCh38
rs760810196 Chr.2: 54255565-54255566 on Build GRCh38
rs371819321 Chr.2: 54255689-54255689 on Build GRCh38
rs779765749 Chr.2: 54255543-54255543 on Build GRCh38
rs1005720134 Chr.2: 54255555-54255555 on Build GRCh38
rs771887695 Chr.2: 54255591-54255591 on Build GRCh38
rs767998320 Chr.2: 54255648-54255648 on Build GRCh38
rs963456682 Chr.2: 54255617-54255618 on Build GRCh38
rs375382594 Chr.2: 54255714-54255714 on Build GRCh38
rs377455363 Chr.2: 54255737-54255737 on Build GRCh38
rs200851714 Chr.2: 54255580-54255580 on Build GRCh38
rs368698929 Chr.2: 54255639-54255639 on Build GRCh38
rs774203944 Chr.2: 54255672-54255672 on Build GRCh38
rs536750975 Chr.2: 54255616-54255616 on Build GRCh38
rs375747246 Chr.2: 54255578-54255578 on Build GRCh38
rs570413794 Chr.2: 54255676-54255676 on Build GRCh38
rs762306404 Chr.2: 54255729-54255729 on Build GRCh38
rs61742696 Chr.2: 54255705-54255705 on Build GRCh38
rs771195486 Chr.2: 54255594-54255594 on Build GRCh38
rs769334448 Chr.2: 54255641-54255641 on Build GRCh38
rs776410736 Chr.2: 54255649-54255649 on Build GRCh38
rs752201041 Chr.2: 54255660-54255660 on Build GRCh38
rs796797230 Chr.2: 54255567-54255572 on Build GRCh38
rs760946195 Chr.2: 54255569-54255569 on Build GRCh38
rs542405286 Chr.2: 54255751-54255751 on Build GRCh38
rs754949900 Chr.2: 54255579-54255580 on Build GRCh38
rs771768568 Chr.2: 54255560-54255560 on Build GRCh38
rs375383770 Chr.2: 54255636-54255636 on Build GRCh38
rs777613409 Chr.2: 54255592-54255592 on Build GRCh38
rs747927664 Chr.2: 54255557-54255557 on Build GRCh38
rs773490596 Chr.2: 54255711-54255711 on Build GRCh38
rs1037864861 Chr.2: 54255541-54255541 on Build GRCh38
rs748048589 Chr.2: 54255589-54255589 on Build GRCh38
rs76397255 Chr.2: 54255579-54255584 on Build GRCh38
rs754017156 Chr.2: 54255566-54255567 on Build GRCh38
rs775019038 Chr.2: 54255644-54255644 on Build GRCh38
rs757079209 Chr.2: 54255633-54255633 on Build GRCh38
rs752010070 Chr.2: 54255597-54255597 on Build GRCh38
rs779914539 Chr.2: 54255570-54255575 on Build GRCh38
rs746095026 Chr.2: 54255685-54255685 on Build GRCh38
rs781018740 Chr.2: 54255684-54255684 on Build GRCh38
rs759576559 Chr.2: 54255595-54255595 on Build GRCh38
rs369009260 Chr.2: 54255724-54255724 on Build GRCh38
rs762227245 Chr.2: 54255647-54255647 on Build GRCh38
rs763983195 Chr.2: 54255674-54255674 on Build GRCh38
rs1022215865 Chr.2: 54255673-54255673 on Build GRCh38
rs776734397 Chr.2: 54255572-54255572 on Build GRCh38
rs375133201 Chr.2: 54255691-54255691 on Build GRCh38
rs757951458 Chr.2: 54255669-54255669 on Build GRCh38
rs774931245 Chr.2: 54255703-54255703 on Build GRCh38
rs370834903 Chr.2: 54255556-54255556 on Build GRCh38
rs891530256 Chr.2: 54255678-54255678 on Build GRCh38
rs750236892 Chr.2: 54255752-54255752 on Build GRCh38
rs760017792 Chr.2: 54255574-54255574 on Build GRCh38
rs747203329 Chr.2: 54255593-54255593 on Build GRCh38
rs371422603 Chr.2: 54255635-54255635 on Build GRCh38
rs755895568 Chr.2: 54255577-54255577 on Build GRCh38
rs907954546 Chr.2: 54255603-54255603 on Build GRCh38
rs756983358 Chr.2: 54255680-54255680 on Build GRCh38
rs973521434 Chr.2: 54255631-54255631 on Build GRCh38
rs754467247 Chr.2: 54255585-54255585 on Build GRCh38
rs750084547 Chr.2: 54255576-54255576 on Build GRCh38
rs548021910 Chr.2: 54255598-54255598 on Build GRCh38
rs901605541 Chr.2: 54255543-54255560 on Build GRCh38
rs890061125 Chr.2: 54255548-54255548 on Build GRCh38
rs756328873 Chr.2: 54255687-54255687 on Build GRCh38
rs369616793 Chr.2: 54255702-54255702 on Build GRCh38
rs747714322 Chr.2: 54255661-54255663 on Build GRCh38
rs766079490 Chr.2: 54255599-54255599 on Build GRCh38
rs370598518 Chr.2: 54255588-54255588 on Build GRCh38
rs773427483 Chr.2: 54255567-54255567 on Build GRCh38
rs1042882931 Chr.2: 54255712-54255712 on Build GRCh38
rs772561126 Chr.2: 54255707-54255707 on Build GRCh38
rs367701165 Chr.2: 54255627-54255627 on Build GRCh38
rs532273037 Chr.2: 54255542-54255542 on Build GRCh38
rs768677445 Chr.2: 54255547-54255547 on Build GRCh38
rs746760341 Chr.2: 54255617-54255617 on Build GRCh38
rs759178976 Chr.2: 54255650-54255650 on Build GRCh38
rs781086080 Chr.2: 54255591-54255592 on Build GRCh38
rs202006037 Chr.2: 54255596-54255596 on Build GRCh38

View Full Product Details