Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 276702-276935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf96
Entrez Gene ID
140680
Gene Name
chromosome 20 open reading frame 96
Gene Aliases
dJ1103G7.2
UniGene
Hs.348112
Gene Chromosome Location
Chr.20: 270863-290778 on Build GRCh38
Transcripts
NM_080571.1 NM_153269.2 XM_006723543.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7224521 c.713T>A p.L238Q Chr.20: 276818-276818 on Build GRCh38
3963424 c.782T>A p.L261Q Chr.20: 276818-276818 on Build GRCh38
6615604 c.831C>T p.T277T Chr.20: 276874-276874 on Build GRCh38
5527709 c.711C>T p.F237F Chr.20: 276820-276820 on Build GRCh38
6230170 c.743G>A p.R248H Chr.20: 276788-276788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541368648 Chr.20: 276794-276794 on Build GRCh38
rs750242886 Chr.20: 276809-276809 on Build GRCh38
rs750356412 Chr.20: 276891-276891 on Build GRCh38
rs745537218 Chr.20: 276724-276724 on Build GRCh38
rs754559449 Chr.20: 276820-276820 on Build GRCh38
rs966091631 Chr.20: 276902-276902 on Build GRCh38
rs954904019 Chr.20: 276906-276906 on Build GRCh38
rs772525783 Chr.20: 276843-276843 on Build GRCh38
rs138308456 Chr.20: 276747-276747 on Build GRCh38
rs372862648 Chr.20: 276897-276897 on Build GRCh38
rs748695512 Chr.20: 276839-276839 on Build GRCh38
rs761027689 Chr.20: 276788-276788 on Build GRCh38
rs375941915 Chr.20: 276908-276908 on Build GRCh38
rs957773345 Chr.20: 276814-276814 on Build GRCh38
rs779839054 Chr.20: 276733-276733 on Build GRCh38
rs776799672 Chr.20: 276790-276790 on Build GRCh38
rs748856490 Chr.20: 276734-276734 on Build GRCh38
rs761705990 Chr.20: 276873-276873 on Build GRCh38
rs185436417 Chr.20: 276738-276738 on Build GRCh38
rs767403827 Chr.20: 276886-276886 on Build GRCh38
rs113722416 Chr.20: 276870-276870 on Build GRCh38
rs138663865 Chr.20: 276795-276795 on Build GRCh38
rs773384373 Chr.20: 276764-276764 on Build GRCh38
rs769920971 Chr.20: 276875-276875 on Build GRCh38
rs763074477 Chr.20: 276883-276883 on Build GRCh38
rs562163373 Chr.20: 276728-276728 on Build GRCh38
rs754524136 Chr.20: 276910-276910 on Build GRCh38
rs766650088 Chr.20: 276789-276789 on Build GRCh38
rs747411459 Chr.20: 276863-276863 on Build GRCh38
rs767485299 Chr.20: 276808-276808 on Build GRCh38
rs757074568 Chr.20: 276803-276803 on Build GRCh38
rs755952776 Chr.20: 276811-276811 on Build GRCh38
rs779892545 Chr.20: 276815-276815 on Build GRCh38
rs989299616 Chr.20: 276842-276842 on Build GRCh38
rs149323011 Chr.20: 276799-276799 on Build GRCh38
rs1002362911 Chr.20: 276802-276802 on Build GRCh38
rs367592807 Chr.20: 276763-276763 on Build GRCh38
rs748555371 Chr.20: 276758-276758 on Build GRCh38
rs778334608 Chr.20: 276845-276845 on Build GRCh38
rs778232002 Chr.20: 276838-276838 on Build GRCh38
rs771240781 Chr.20: 276869-276869 on Build GRCh38
rs753504063 Chr.20: 276817-276817 on Build GRCh38

View Full Product Details