Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75240132-75240393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7A2
Entrez Gene ID
1347
Gene Name
cytochrome c oxidase subunit 7A2
Gene Aliases
COX7AL, COX7AL1, COXVIIAL, COXVIIa-L, VIIAL
UniGene
Hs.70312
Gene Chromosome Location
Chr.6: 75237675-75243928 on Build GRCh38
Transcripts
NM_001865.3 NR_029466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4006555 c.254T>C p.L85P Chr.6: 75240336-75240336 on Build GRCh38
5996777 c.158T>G p.L53R Chr.6: 75240336-75240336 on Build GRCh38
1202110 c.120A>C p.E40D Chr.6: 75240374-75240374 on Build GRCh38
3629872 c.252C>T p.A84A Chr.6: 75240338-75240338 on Build GRCh38
6310568 c.109-8A>T p.? Chr.6: 75240393-75240393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890804423 Chr.6: 75240241-75240241 on Build GRCh38
rs959036597 Chr.6: 75240209-75240209 on Build GRCh38
rs11538565 Chr.6: 75240346-75240346 on Build GRCh38
rs528461492 Chr.6: 75240301-75240301 on Build GRCh38
rs752674209 Chr.6: 75240225-75240225 on Build GRCh38
rs553282690 Chr.6: 75240290-75240290 on Build GRCh38
rs777325624 Chr.6: 75240282-75240282 on Build GRCh38
rs1035185960 Chr.6: 75240196-75240196 on Build GRCh38
rs1011279470 Chr.6: 75240314-75240314 on Build GRCh38
rs4708176 Chr.6: 75240180-75240180 on Build GRCh38
rs115820973 Chr.6: 75240340-75240340 on Build GRCh38
rs1050995023 Chr.6: 75240226-75240226 on Build GRCh38
rs368893887 Chr.6: 75240334-75240334 on Build GRCh38
rs754814290 Chr.6: 75240352-75240352 on Build GRCh38
rs778640515 Chr.6: 75240182-75240182 on Build GRCh38
rs967922184 Chr.6: 75240261-75240261 on Build GRCh38
rs537553690 Chr.6: 75240176-75240176 on Build GRCh38
rs567341084 Chr.6: 75240187-75240187 on Build GRCh38
rs892923172 Chr.6: 75240288-75240288 on Build GRCh38
rs752771051 Chr.6: 75240278-75240278 on Build GRCh38
rs754053902 Chr.6: 75240331-75240331 on Build GRCh38
rs371290408 Chr.6: 75240363-75240363 on Build GRCh38
rs1022145680 Chr.6: 75240259-75240259 on Build GRCh38
rs547158921 Chr.6: 75240353-75240353 on Build GRCh38
rs187541079 Chr.6: 75240258-75240258 on Build GRCh38
rs760029628 Chr.6: 75240249-75240249 on Build GRCh38
rs374450683 Chr.6: 75240291-75240291 on Build GRCh38
rs370234188 Chr.6: 75240174-75240174 on Build GRCh38
rs867838661 Chr.6: 75240322-75240322 on Build GRCh38
rs918403353 Chr.6: 75240219-75240219 on Build GRCh38
rs764374816 Chr.6: 75240319-75240319 on Build GRCh38
rs150749777 Chr.6: 75240332-75240332 on Build GRCh38

View Full Product Details