Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79784616-79784841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX2
Entrez Gene ID
84733
Gene Name
chromobox 2
Gene Aliases
CDCA6, M33, SRXY5
UniGene
Hs.368410
Gene Chromosome Location
Chr.17: 79776254-79787650 on Build GRCh38
Transcripts
NM_005189.2 XM_011525382.1 XM_011525383.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4515621 c.1213_1214CC>TT p.P405F Chr.17: 79784656-79784657 on Build GRCh38
6230979 c.1355C>T p.A452V Chr.17: 79784798-79784798 on Build GRCh38
2802777 c.1376G>A p.S459N Chr.17: 79784819-79784819 on Build GRCh38
1303437 c.1371G>A p.E457E Chr.17: 79784814-79784814 on Build GRCh38
1386945 c.1397G>A p.S466N Chr.17: 79784840-79784840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782023433 Chr.17: 79784713-79784713 on Build GRCh38
rs148521974 Chr.17: 79784820-79784820 on Build GRCh38
rs943462815 Chr.17: 79784664-79784664 on Build GRCh38
rs782534826 Chr.17: 79784738-79784738 on Build GRCh38
rs377709788 Chr.17: 79784639-79784639 on Build GRCh38
rs893794222 Chr.17: 79784680-79784680 on Build GRCh38
rs121908256 Chr.17: 79784771-79784771 on Build GRCh38
rs782450964 Chr.17: 79784745-79784745 on Build GRCh38
rs371067953 Chr.17: 79784653-79784653 on Build GRCh38
rs150127376 Chr.17: 79784643-79784643 on Build GRCh38
rs782792566 Chr.17: 79784644-79784644 on Build GRCh38
rs782480937 Chr.17: 79784733-79784733 on Build GRCh38
rs141814853 Chr.17: 79784702-79784702 on Build GRCh38
rs530910308 Chr.17: 79784703-79784703 on Build GRCh38
rs781817531 Chr.17: 79784726-79784726 on Build GRCh38
rs138828014 Chr.17: 79784781-79784781 on Build GRCh38
rs929428272 Chr.17: 79784649-79784649 on Build GRCh38
rs370780535 Chr.17: 79784748-79784748 on Build GRCh38
rs973656706 Chr.17: 79784801-79784801 on Build GRCh38
rs781924919 Chr.17: 79784766-79784766 on Build GRCh38
rs201172710 Chr.17: 79784770-79784770 on Build GRCh38
rs781874682 Chr.17: 79784737-79784737 on Build GRCh38
rs782785209 Chr.17: 79784710-79784710 on Build GRCh38
rs370084807 Chr.17: 79784812-79784812 on Build GRCh38
rs199566347 Chr.17: 79784799-79784799 on Build GRCh38
rs782126290 Chr.17: 79784709-79784709 on Build GRCh38
rs782732296 Chr.17: 79784791-79784791 on Build GRCh38
rs373628706 Chr.17: 79784708-79784708 on Build GRCh38
rs781907844 Chr.17: 79784651-79784651 on Build GRCh38
rs76915888 Chr.17: 79784798-79784798 on Build GRCh38
rs782670771 Chr.17: 79784741-79784741 on Build GRCh38
rs782287402 Chr.17: 79784756-79784756 on Build GRCh38
rs141949674 Chr.17: 79784787-79784787 on Build GRCh38
rs532091014 Chr.17: 79784757-79784757 on Build GRCh38
rs782597003 Chr.17: 79784661-79784661 on Build GRCh38
rs781922744 Chr.17: 79784693-79784693 on Build GRCh38
rs781861149 Chr.17: 79784640-79784640 on Build GRCh38
rs568793911 Chr.17: 79784792-79784792 on Build GRCh38
rs564660447 Chr.17: 79784687-79784687 on Build GRCh38
rs3751956 Chr.17: 79784685-79784685 on Build GRCh38
rs782222190 Chr.17: 79784662-79784662 on Build GRCh38
rs781978951 Chr.17: 79784774-79784774 on Build GRCh38
rs781786898 Chr.17: 79784743-79784743 on Build GRCh38
rs373878749 Chr.17: 79784782-79784782 on Build GRCh38
rs187278438 Chr.17: 79784675-79784675 on Build GRCh38
rs145541835 Chr.17: 79784712-79784712 on Build GRCh38
rs781967736 Chr.17: 79784724-79784724 on Build GRCh38
rs782088256 Chr.17: 79784768-79784768 on Build GRCh38
rs782755628 Chr.17: 79784734-79784734 on Build GRCh38
rs782657688 Chr.17: 79784755-79784755 on Build GRCh38
rs782650777 Chr.17: 79784684-79784684 on Build GRCh38

View Full Product Details