Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 45428547-45428806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5
Entrez Gene ID
240
Gene Name
arachidonate 5-lipoxygenase
Gene Aliases
5-LO, 5-LOX, 5LPG, LOG5
UniGene
Hs.89499
Gene Chromosome Location
Chr.10: 45374166-45446121 on Build GRCh38
Transcripts
NM_000698.4 NM_001256153.2 NM_001256154.2 NM_001320861.1 NM_001320862.1 XM_017016012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5589618 c.895G>A p.D299N Chr.10: 45428678-45428678 on Build GRCh38
6424654 c.902G>A p.C301Y Chr.10: 45428685-45428685 on Build GRCh38
458830 c.915C>T p.F305F Chr.10: 45428698-45428698 on Build GRCh38
1347871 c.853G>A p.V285M Chr.10: 45428636-45428636 on Build GRCh38
4838120 c.883G>A p.A295T Chr.10: 45428666-45428666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759600909 Chr.10: 45428728-45428728 on Build GRCh38
rs767991624 Chr.10: 45428636-45428636 on Build GRCh38
rs201471525 Chr.10: 45428688-45428688 on Build GRCh38
rs373501906 Chr.10: 45428574-45428574 on Build GRCh38
rs779835358 Chr.10: 45428754-45428754 on Build GRCh38
rs768081589 Chr.10: 45428729-45428729 on Build GRCh38
rs28395872 Chr.10: 45428705-45428705 on Build GRCh38
rs768768474 Chr.10: 45428671-45428671 on Build GRCh38
rs199926259 Chr.10: 45428692-45428692 on Build GRCh38
rs776702675 Chr.10: 45428680-45428680 on Build GRCh38
rs755519785 Chr.10: 45428625-45428625 on Build GRCh38
rs758032522 Chr.10: 45428753-45428753 on Build GRCh38
rs369157831 Chr.10: 45428638-45428638 on Build GRCh38
rs900958723 Chr.10: 45428651-45428651 on Build GRCh38
rs376303888 Chr.10: 45428740-45428740 on Build GRCh38
rs761122303 Chr.10: 45428732-45428732 on Build GRCh38
rs1053114916 Chr.10: 45428606-45428606 on Build GRCh38
rs1565096 Chr.10: 45428575-45428575 on Build GRCh38
rs150216824 Chr.10: 45428660-45428660 on Build GRCh38
rs773509478 Chr.10: 45428702-45428702 on Build GRCh38
rs756403009 Chr.10: 45428639-45428639 on Build GRCh38
rs534018579 Chr.10: 45428731-45428731 on Build GRCh38
rs762238091 Chr.10: 45428773-45428773 on Build GRCh38
rs762204941 Chr.10: 45428749-45428749 on Build GRCh38
rs45598533 Chr.10: 45428695-45428695 on Build GRCh38
rs754198918 Chr.10: 45428748-45428748 on Build GRCh38
rs535808410 Chr.10: 45428663-45428663 on Build GRCh38
rs370294843 Chr.10: 45428664-45428664 on Build GRCh38
rs1002083806 Chr.10: 45428772-45428772 on Build GRCh38
rs768038790 Chr.10: 45428775-45428775 on Build GRCh38
rs369666935 Chr.10: 45428568-45428568 on Build GRCh38
rs752091985 Chr.10: 45428579-45428579 on Build GRCh38
rs769421758 Chr.10: 45428699-45428699 on Build GRCh38
rs757957452 Chr.10: 45428650-45428650 on Build GRCh38
rs745461924 Chr.10: 45428644-45428644 on Build GRCh38

View Full Product Details