Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 16514473-16514746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QL3
Entrez Gene ID
389941
Gene Name
complement C1q like 3
Gene Aliases
C1QTNF13, C1ql, CTRP13, K100
UniGene
Hs.676792
Gene Chromosome Location
Chr.10: 16513743-16522005 on Build GRCh38
Transcripts
NM_001010908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7216408 c.589-4A>G p.? Chr.10: 16514711-16514711 on Build GRCh38
465497 c.592C>T p.R198C Chr.10: 16514704-16514704 on Build GRCh38
2131520 c.630C>T p.Y210Y Chr.10: 16514666-16514666 on Build GRCh38
6013765 c.669G>A p.P223P Chr.10: 16514627-16514627 on Build GRCh38
2131518 c.657T>C p.L219L Chr.10: 16514639-16514639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775095801 Chr.10: 16514628-16514628 on Build GRCh38
rs774959199 Chr.10: 16514558-16514558 on Build GRCh38
rs777367197 Chr.10: 16514531-16514531 on Build GRCh38
rs748536717 Chr.10: 16514645-16514645 on Build GRCh38
rs758395688 Chr.10: 16514526-16514526 on Build GRCh38
rs773433390 Chr.10: 16514687-16514687 on Build GRCh38
rs1029691573 Chr.10: 16514614-16514614 on Build GRCh38
rs769237834 Chr.10: 16514617-16514617 on Build GRCh38
rs766554141 Chr.10: 16514705-16514705 on Build GRCh38
rs772645414 Chr.10: 16514666-16514666 on Build GRCh38
rs138237084 Chr.10: 16514597-16514597 on Build GRCh38
rs377239962 Chr.10: 16514511-16514511 on Build GRCh38
rs756959476 Chr.10: 16514563-16514563 on Build GRCh38
rs7909832 Chr.10: 16514711-16514711 on Build GRCh38
rs762420617 Chr.10: 16514710-16514710 on Build GRCh38
rs954969613 Chr.10: 16514678-16514678 on Build GRCh38
rs374984474 Chr.10: 16514501-16514501 on Build GRCh38
rs775177429 Chr.10: 16514703-16514704 on Build GRCh38
rs368514858 Chr.10: 16514504-16514504 on Build GRCh38
rs746816359 Chr.10: 16514536-16514536 on Build GRCh38
rs372477560 Chr.10: 16514613-16514613 on Build GRCh38
rs752532411 Chr.10: 16514518-16514518 on Build GRCh38
rs765195473 Chr.10: 16514714-16514714 on Build GRCh38
rs114662300 Chr.10: 16514512-16514512 on Build GRCh38
rs759772609 Chr.10: 16514712-16514712 on Build GRCh38
rs769354377 Chr.10: 16514516-16514517 on Build GRCh38
rs147489406 Chr.10: 16514546-16514546 on Build GRCh38
rs761091339 Chr.10: 16514704-16514704 on Build GRCh38
rs1008332125 Chr.10: 16514692-16514692 on Build GRCh38

View Full Product Details