Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152863527-152863776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP1
Entrez Gene ID
27236
Gene Name
ADP ribosylation factor interacting protein 1
Gene Aliases
HSU52521
UniGene
Hs.416089
Gene Chromosome Location
Chr.4: 152779937-152911912 on Build GRCh38
Transcripts
NM_001025593.2 NM_001025595.2 NM_001287431.1 NM_001287432.1 NM_001287433.1 NM_014447.3 XM_011531868.2 XM_011531869.2 XM_011531870.2 XM_011531871.2 XM_011531872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4470744 c.167C>T p.T56I Chr.4: 152863679-152863679 on Build GRCh38
3365525 c.107C>A p.S36* Chr.4: 152863619-152863619 on Build GRCh38
4532801 c.191G>C p.G64A Chr.4: 152863703-152863703 on Build GRCh38
5883953 c.149C>G p.S50C Chr.4: 152863661-152863661 on Build GRCh38
5883954 c.149C>G p.S50C Chr.4: 152863661-152863661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770036152 Chr.4: 152863736-152863736 on Build GRCh38
rs994892017 Chr.4: 152863686-152863686 on Build GRCh38
rs747539544 Chr.4: 152863676-152863676 on Build GRCh38
rs753497476 Chr.4: 152863621-152863621 on Build GRCh38
rs141673805 Chr.4: 152863709-152863709 on Build GRCh38
rs534134939 Chr.4: 152863743-152863743 on Build GRCh38
rs138793616 Chr.4: 152863564-152863565 on Build GRCh38
rs773304818 Chr.4: 152863575-152863575 on Build GRCh38
rs758904927 Chr.4: 152863666-152863666 on Build GRCh38
rs536318352 Chr.4: 152863705-152863705 on Build GRCh38
rs775089939 Chr.4: 152863592-152863592 on Build GRCh38
rs759429581 Chr.4: 152863581-152863581 on Build GRCh38
rs960579003 Chr.4: 152863558-152863558 on Build GRCh38
rs141072061 Chr.4: 152863571-152863572 on Build GRCh38
rs568899855 Chr.4: 152863574-152863574 on Build GRCh38
rs1008893153 Chr.4: 152863722-152863722 on Build GRCh38
rs767447500 Chr.4: 152863589-152863589 on Build GRCh38
rs1055259739 Chr.4: 152863633-152863633 on Build GRCh38
rs773210934 Chr.4: 152863740-152863740 on Build GRCh38
rs371942166 Chr.4: 152863694-152863694 on Build GRCh38
rs1002877046 Chr.4: 152863567-152863567 on Build GRCh38
rs1043130586 Chr.4: 152863706-152863706 on Build GRCh38
rs1019373469 Chr.4: 152863733-152863733 on Build GRCh38
rs965287491 Chr.4: 152863739-152863739 on Build GRCh38
rs368596152 Chr.4: 152863639-152863639 on Build GRCh38
rs768964016 Chr.4: 152863562-152863562 on Build GRCh38
rs374481689 Chr.4: 152863601-152863601 on Build GRCh38
rs749980564 Chr.4: 152863653-152863653 on Build GRCh38
rs780601305 Chr.4: 152863673-152863673 on Build GRCh38
rs747400075 Chr.4: 152863559-152863559 on Build GRCh38
rs567015324 Chr.4: 152863723-152863723 on Build GRCh38
rs757017032 Chr.4: 152863624-152863624 on Build GRCh38
rs147195311 Chr.4: 152863691-152863691 on Build GRCh38
rs777993394 Chr.4: 152863662-152863662 on Build GRCh38
rs762065971 Chr.4: 152863565-152863565 on Build GRCh38
rs763860418 Chr.4: 152863620-152863620 on Build GRCh38
rs749541326 Chr.4: 152863737-152863737 on Build GRCh38

View Full Product Details