Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38673856-38674105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf96
Entrez Gene ID
100170841
Gene Name
chromosome 17 open reading frame 96
Gene Aliases
PRR28
UniGene
Hs.445574
Gene Chromosome Location
Chr.17: 38671703-38674934 on Build GRCh38
Transcripts
NM_001130677.1

SNP Detail

SNP ID Chromosome Location
rs1012973530 Chr.17: 38673922-38673922 on Build GRCh38
rs533951939 Chr.17: 38674067-38674067 on Build GRCh38
rs775620836 Chr.17: 38674042-38674042 on Build GRCh38
rs568178963 Chr.17: 38673984-38673984 on Build GRCh38
rs1022641882 Chr.17: 38673978-38673978 on Build GRCh38
rs1037057562 Chr.17: 38674046-38674046 on Build GRCh38
rs922544053 Chr.17: 38673927-38673927 on Build GRCh38
rs955850496 Chr.17: 38674041-38674041 on Build GRCh38
rs954174581 Chr.17: 38674069-38674069 on Build GRCh38
rs1031633291 Chr.17: 38674036-38674036 on Build GRCh38
rs968469041 Chr.17: 38673881-38673881 on Build GRCh38
rs777935504 Chr.17: 38673923-38673923 on Build GRCh38
rs924297682 Chr.17: 38673886-38673886 on Build GRCh38
rs772276333 Chr.17: 38673880-38673880 on Build GRCh38
rs1056794110 Chr.17: 38673893-38673893 on Build GRCh38
rs761052437 Chr.17: 38674057-38674057 on Build GRCh38
rs777192794 Chr.17: 38673952-38673952 on Build GRCh38
rs978442534 Chr.17: 38673882-38673882 on Build GRCh38
rs932482365 Chr.17: 38673940-38673940 on Build GRCh38
rs1000118139 Chr.17: 38674011-38674011 on Build GRCh38
rs964240546 Chr.17: 38674062-38674062 on Build GRCh38
rs895589818 Chr.17: 38673902-38673902 on Build GRCh38
rs909967648 Chr.17: 38674006-38674006 on Build GRCh38

View Full Product Details