Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71605570-71605835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACRC
Entrez Gene ID
93953
Gene Name
acidic repeat containing
Gene Aliases
NAAR1
UniGene
Hs.135167
Gene Chromosome Location
Chr.X: 71578024-71613583 on Build GRCh38
Transcripts
NM_052957.4 XM_006724716.3 XM_011531063.2 XM_011531064.1 XM_011531066.2 XM_011531067.2 XM_017029965.1

SNP Detail

SNP ID Chromosome Location
rs750713454 Chr.X: 71605768-71605768 on Build GRCh38
rs756265580 Chr.X: 71605776-71605776 on Build GRCh38
rs779348280 Chr.X: 71605701-71605701 on Build GRCh38
rs757236212 Chr.X: 71605651-71605651 on Build GRCh38
rs374858167 Chr.X: 71605719-71605719 on Build GRCh38
rs760313290 Chr.X: 71605747-71605747 on Build GRCh38
rs772802338 Chr.X: 71605720-71605720 on Build GRCh38
rs768598181 Chr.X: 71605791-71605791 on Build GRCh38
rs1023953182 Chr.X: 71605785-71605785 on Build GRCh38
rs764311492 Chr.X: 71605613-71605613 on Build GRCh38
rs780380763 Chr.X: 71605681-71605681 on Build GRCh38
rs369238580 Chr.X: 71605732-71605732 on Build GRCh38
rs775395731 Chr.X: 71605733-71605733 on Build GRCh38
rs761956360 Chr.X: 71605743-71605743 on Build GRCh38
rs760877582 Chr.X: 71605752-71605752 on Build GRCh38
rs185057741 Chr.X: 71605714-71605714 on Build GRCh38
rs764150065 Chr.X: 71605739-71605739 on Build GRCh38
rs749840112 Chr.X: 71605697-71605697 on Build GRCh38
rs767793682 Chr.X: 71605674-71605674 on Build GRCh38
rs772374706 Chr.X: 71605703-71605703 on Build GRCh38
rs746110671 Chr.X: 71605645-71605645 on Build GRCh38
rs762796801 Chr.X: 71605736-71605736 on Build GRCh38
rs771938104 Chr.X: 71605698-71605698 on Build GRCh38
rs10217999 Chr.X: 71605675-71605675 on Build GRCh38
rs756567517 Chr.X: 71605680-71605680 on Build GRCh38
rs774386539 Chr.X: 71605644-71605645 on Build GRCh38
rs371837884 Chr.X: 71605742-71605742 on Build GRCh38

View Full Product Details