Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115992118-115992343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LVRN
Entrez Gene ID
206338
Gene Name
laeverin
Gene Aliases
APQ, AQPEP, TAQPEP
UniGene
Hs.556880
Gene Chromosome Location
Chr.5: 115962440-116027602 on Build GRCh38
Transcripts
NM_173800.4 XM_011543263.2 XM_011543264.2 XM_017009215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4625385 c.1176A>T p.S392S Chr.5: 115992193-115992193 on Build GRCh38
1671818 c.1253G>C p.G418A Chr.5: 115992270-115992270 on Build GRCh38
6584527 c.1229C>G p.S410C Chr.5: 115992246-115992246 on Build GRCh38
481775 c.1170T>A p.D390E Chr.5: 115992187-115992187 on Build GRCh38
6602646 c.1214A>G p.K405R Chr.5: 115992231-115992231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758853097 Chr.5: 115992152-115992152 on Build GRCh38
rs745429491 Chr.5: 115992173-115992173 on Build GRCh38
rs570395139 Chr.5: 115992190-115992190 on Build GRCh38
rs780934473 Chr.5: 115992237-115992237 on Build GRCh38
rs753977961 Chr.5: 115992204-115992204 on Build GRCh38
rs372326916 Chr.5: 115992247-115992247 on Build GRCh38
rs757594151 Chr.5: 115992236-115992236 on Build GRCh38
rs928823091 Chr.5: 115992183-115992183 on Build GRCh38
rs1019652380 Chr.5: 115992280-115992280 on Build GRCh38
rs139728913 Chr.5: 115992252-115992252 on Build GRCh38
rs778123320 Chr.5: 115992161-115992161 on Build GRCh38
rs553023548 Chr.5: 115992282-115992282 on Build GRCh38
rs764060120 Chr.5: 115992184-115992184 on Build GRCh38
rs369197606 Chr.5: 115992263-115992263 on Build GRCh38
rs745534740 Chr.5: 115992240-115992240 on Build GRCh38
rs765254890 Chr.5: 115992214-115992214 on Build GRCh38
rs770635512 Chr.5: 115992164-115992164 on Build GRCh38
rs889768409 Chr.5: 115992266-115992266 on Build GRCh38
rs761742572 Chr.5: 115992254-115992254 on Build GRCh38
rs763004476 Chr.5: 115992307-115992307 on Build GRCh38
rs370891912 Chr.5: 115992191-115992191 on Build GRCh38
rs757717231 Chr.5: 115992315-115992315 on Build GRCh38
rs752785628 Chr.5: 115992295-115992295 on Build GRCh38
rs765236083 Chr.5: 115992215-115992215 on Build GRCh38
rs149831297 Chr.5: 115992267-115992267 on Build GRCh38
rs776132785 Chr.5: 115992171-115992171 on Build GRCh38
rs764631832 Chr.5: 115992310-115992310 on Build GRCh38
rs748807072 Chr.5: 115992249-115992249 on Build GRCh38
rs368373698 Chr.5: 115992174-115992174 on Build GRCh38
rs774358639 Chr.5: 115992187-115992187 on Build GRCh38
rs752026649 Chr.5: 115992312-115992312 on Build GRCh38
rs747305567 Chr.5: 115992162-115992162 on Build GRCh38
rs752763978 Chr.5: 115992217-115992217 on Build GRCh38
rs762817050 Chr.5: 115992182-115992182 on Build GRCh38
rs769435558 Chr.5: 115992246-115992246 on Build GRCh38
rs763081983 Chr.5: 115992309-115992310 on Build GRCh38
rs1033871119 Chr.5: 115992193-115992193 on Build GRCh38
rs775920992 Chr.5: 115992250-115992250 on Build GRCh38
rs376904306 Chr.5: 115992262-115992262 on Build GRCh38
rs755188589 Chr.5: 115992209-115992209 on Build GRCh38
rs775565481 Chr.5: 115992295-115992296 on Build GRCh38
rs747430037 Chr.5: 115992235-115992235 on Build GRCh38
rs965392012 Chr.5: 115992308-115992308 on Build GRCh38
rs758910161 Chr.5: 115992222-115992222 on Build GRCh38
rs775021811 Chr.5: 115992177-115992177 on Build GRCh38

View Full Product Details