Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165493066-165493313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPE
Entrez Gene ID
1363
Gene Name
carboxypeptidase E
Gene Aliases
CPH
UniGene
Hs.75360
Gene Chromosome Location
Chr.4: 165378942-165498547 on Build GRCh38
Transcripts
NM_001873.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5437690 c.1139T>C p.V380A Chr.4: 165493196-165493196 on Build GRCh38
5918375 c.1190G>A p.G397E Chr.4: 165493247-165493247 on Build GRCh38
5687964 c.1213G>C p.A405P Chr.4: 165493270-165493270 on Build GRCh38
4459926 c.1147C>T p.L383F Chr.4: 165493204-165493204 on Build GRCh38
6049920 c.1120C>A p.R374R Chr.4: 165493177-165493177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777834236 Chr.4: 165493165-165493165 on Build GRCh38
rs748297557 Chr.4: 165493159-165493159 on Build GRCh38
rs769928218 Chr.4: 165493162-165493162 on Build GRCh38
rs956945391 Chr.4: 165493209-165493209 on Build GRCh38
rs144687736 Chr.4: 165493234-165493234 on Build GRCh38
rs763836636 Chr.4: 165493128-165493128 on Build GRCh38
rs935207686 Chr.4: 165493115-165493115 on Build GRCh38
rs200971710 Chr.4: 165493145-165493145 on Build GRCh38
rs756948371 Chr.4: 165493276-165493276 on Build GRCh38
rs776399312 Chr.4: 165493218-165493218 on Build GRCh38
rs772865461 Chr.4: 165493257-165493257 on Build GRCh38
rs780653193 Chr.4: 165493149-165493149 on Build GRCh38
rs558171413 Chr.4: 165493287-165493287 on Build GRCh38
rs761431084 Chr.4: 165493129-165493129 on Build GRCh38
rs375881004 Chr.4: 165493265-165493265 on Build GRCh38
rs1001165792 Chr.4: 165493160-165493160 on Build GRCh38
rs140738622 Chr.4: 165493195-165493195 on Build GRCh38
rs764830554 Chr.4: 165493133-165493133 on Build GRCh38
rs1036060971 Chr.4: 165493232-165493232 on Build GRCh38
rs764918504 Chr.4: 165493238-165493238 on Build GRCh38
rs557463203 Chr.4: 165493153-165493153 on Build GRCh38
rs1002974047 Chr.4: 165493213-165493213 on Build GRCh38
rs749306072 Chr.4: 165493180-165493180 on Build GRCh38
rs746859518 Chr.4: 165493210-165493210 on Build GRCh38
rs768310022 Chr.4: 165493212-165493212 on Build GRCh38
rs368423308 Chr.4: 165493122-165493122 on Build GRCh38
rs568249212 Chr.4: 165493144-165493144 on Build GRCh38
rs767026740 Chr.4: 165493280-165493280 on Build GRCh38
rs1012109303 Chr.4: 165493141-165493141 on Build GRCh38
rs775367854 Chr.4: 165493120-165493120 on Build GRCh38
rs781518085 Chr.4: 165493154-165493154 on Build GRCh38
rs988153629 Chr.4: 165493224-165493224 on Build GRCh38
rs767767805 Chr.4: 165493288-165493288 on Build GRCh38
rs994819145 Chr.4: 165493261-165493261 on Build GRCh38
rs751977465 Chr.4: 165493150-165493150 on Build GRCh38
rs868366203 Chr.4: 165493255-165493255 on Build GRCh38
rs751980730 Chr.4: 165493283-165493283 on Build GRCh38
rs775247571 Chr.4: 165493200-165493200 on Build GRCh38

View Full Product Details