Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45480144-45480397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG3
Entrez Gene ID
83548
Gene Name
component of oligomeric golgi complex 3
Gene Aliases
SEC34
UniGene
Hs.507948
Gene Chromosome Location
Chr.13: 45464895-45536698 on Build GRCh38
Transcripts
NM_031431.3 XM_011535266.1 XM_011535267.2 XM_017020790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1135499 c.549+2T>C p.? Chr.13: 45480292-45480292 on Build GRCh38
7179228 c.531A>G p.E177E Chr.13: 45480272-45480272 on Build GRCh38
6880612 c.481C>A p.Q161K Chr.13: 45480222-45480222 on Build GRCh38
1941557 c.438T>C p.N146N Chr.13: 45480179-45480179 on Build GRCh38
6588458 c.412C>G p.Q138E Chr.13: 45480153-45480153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041604141 Chr.13: 45480174-45480174 on Build GRCh38
rs753218301 Chr.13: 45480242-45480242 on Build GRCh38
rs1011124614 Chr.13: 45480272-45480272 on Build GRCh38
rs938490890 Chr.13: 45480236-45480236 on Build GRCh38
rs745326793 Chr.13: 45480308-45480308 on Build GRCh38
rs756904905 Chr.13: 45480259-45480259 on Build GRCh38
rs553387280 Chr.13: 45480348-45480348 on Build GRCh38
rs780719393 Chr.13: 45480301-45480301 on Build GRCh38
rs1056968644 Chr.13: 45480240-45480240 on Build GRCh38
rs1008965274 Chr.13: 45480356-45480356 on Build GRCh38
rs755671209 Chr.13: 45480317-45480317 on Build GRCh38
rs376780940 Chr.13: 45480238-45480238 on Build GRCh38
rs1026542941 Chr.13: 45480344-45480344 on Build GRCh38
rs111573190 Chr.13: 45480340-45480340 on Build GRCh38
rs749241135 Chr.13: 45480324-45480324 on Build GRCh38
rs373281254 Chr.13: 45480171-45480171 on Build GRCh38
rs762784820 Chr.13: 45480185-45480185 on Build GRCh38
rs761045385 Chr.13: 45480203-45480203 on Build GRCh38
rs753905944 Chr.13: 45480222-45480222 on Build GRCh38
rs778180574 Chr.13: 45480254-45480254 on Build GRCh38
rs79928919 Chr.13: 45480223-45480223 on Build GRCh38
rs773500342 Chr.13: 45480199-45480199 on Build GRCh38
rs768584251 Chr.13: 45480190-45480190 on Build GRCh38
rs755963791 Chr.13: 45480304-45480305 on Build GRCh38
rs954984735 Chr.13: 45480352-45480352 on Build GRCh38
rs889259706 Chr.13: 45480345-45480345 on Build GRCh38
rs1016062245 Chr.13: 45480261-45480261 on Build GRCh38
rs779797162 Chr.13: 45480323-45480323 on Build GRCh38
rs766461048 Chr.13: 45480221-45480221 on Build GRCh38
rs202080854 Chr.13: 45480302-45480302 on Build GRCh38
rs774336481 Chr.13: 45480333-45480333 on Build GRCh38
rs775527182 Chr.13: 45480178-45480178 on Build GRCh38
rs764018028 Chr.13: 45480326-45480327 on Build GRCh38
rs371004147 Chr.13: 45480257-45480257 on Build GRCh38
rs894183782 Chr.13: 45480246-45480246 on Build GRCh38
rs141980886 Chr.13: 45480325-45480325 on Build GRCh38

View Full Product Details