Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87409635-87409908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP206
Entrez Gene ID
154313
Gene Name
cilia and flagella associated protein 206
Gene Aliases
C6orf165, dJ382I10.1
UniGene
Hs.82921
Gene Chromosome Location
Chr.6: 87407972-87464473 on Build GRCh38
Transcripts
NM_001031743.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1622003 c.16G>A p.A6T Chr.6: 87409855-87409855 on Build GRCh38
3922231 c.44G>A p.R15Q Chr.6: 87409883-87409883 on Build GRCh38
3922230 c.44G>A p.R15Q Chr.6: 87409883-87409883 on Build GRCh38
3177719 c.32G>T p.R11M Chr.6: 87409871-87409871 on Build GRCh38
6488984 c.69C>A p.A23A Chr.6: 87409908-87409908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771100334 Chr.6: 87409794-87409794 on Build GRCh38
rs777296517 Chr.6: 87409829-87409829 on Build GRCh38
rs772499613 Chr.6: 87409857-87409857 on Build GRCh38
rs955605594 Chr.6: 87409672-87409672 on Build GRCh38
rs776515922 Chr.6: 87409802-87409802 on Build GRCh38
rs774204452 Chr.6: 87409883-87409883 on Build GRCh38
rs941219282 Chr.6: 87409727-87409727 on Build GRCh38
rs776955208 Chr.6: 87409875-87409875 on Build GRCh38
rs780296013 Chr.6: 87409863-87409863 on Build GRCh38
rs201057926 Chr.6: 87409851-87409851 on Build GRCh38
rs775776886 Chr.6: 87409790-87409790 on Build GRCh38
rs373302872 Chr.6: 87409824-87409824 on Build GRCh38
rs1049165990 Chr.6: 87409776-87409776 on Build GRCh38
rs150062349 Chr.6: 87409882-87409882 on Build GRCh38
rs200029568 Chr.6: 87409811-87409811 on Build GRCh38
rs764413218 Chr.6: 87409818-87409821 on Build GRCh38
rs918486882 Chr.6: 87409738-87409738 on Build GRCh38
rs531012169 Chr.6: 87409856-87409856 on Build GRCh38
rs187143045 Chr.6: 87409745-87409745 on Build GRCh38
rs948157109 Chr.6: 87409776-87409776 on Build GRCh38
rs753255751 Chr.6: 87409833-87409833 on Build GRCh38
rs760036492 Chr.6: 87409803-87409804 on Build GRCh38
rs960205752 Chr.6: 87409685-87409685 on Build GRCh38
rs776599912 Chr.6: 87409810-87409810 on Build GRCh38
rs766398566 Chr.6: 87409837-87409837 on Build GRCh38
rs543634123 Chr.6: 87409782-87409782 on Build GRCh38
rs377497011 Chr.6: 87409867-87409867 on Build GRCh38
rs755712676 Chr.6: 87409885-87409891 on Build GRCh38
rs985519214 Chr.6: 87409684-87409684 on Build GRCh38
rs796361714 Chr.6: 87409818-87409818 on Build GRCh38
rs752332482 Chr.6: 87409821-87409821 on Build GRCh38
rs11350856 Chr.6: 87409811-87409811 on Build GRCh38
rs909260143 Chr.6: 87409785-87409785 on Build GRCh38
rs911028065 Chr.6: 87409687-87409687 on Build GRCh38
rs767217835 Chr.6: 87409819-87409819 on Build GRCh38
rs973901111 Chr.6: 87409731-87409731 on Build GRCh38
rs76198921 Chr.6: 87409845-87409845 on Build GRCh38
rs768728371 Chr.6: 87409804-87409804 on Build GRCh38
rs763354721 Chr.6: 87409810-87409811 on Build GRCh38
rs530151510 Chr.6: 87409816-87409816 on Build GRCh38
rs768796334 Chr.6: 87409868-87409868 on Build GRCh38
rs760811736 Chr.6: 87409802-87409802 on Build GRCh38
rs751553291 Chr.6: 87409812-87409812 on Build GRCh38
rs761270055 Chr.6: 87409804-87409804 on Build GRCh38
rs998551141 Chr.6: 87409874-87409874 on Build GRCh38
rs550122833 Chr.6: 87409819-87409821 on Build GRCh38
rs991365455 Chr.6: 87409697-87409697 on Build GRCh38

View Full Product Details