Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58306247-58306520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPOAP1
Entrez Gene ID
9256
Gene Name
TSPO associated protein 1
Gene Aliases
BZRAP1, PBR-IP, PRAX-1, PRAX1, RIM-BP1, RIMBP1
UniGene
Hs.112499
Gene Chromosome Location
Chr.17: 58301227-58329030 on Build GRCh38
Transcripts
NM_001261835.1 NM_004758.3 NM_024418.2 XM_005257807.2 XM_006722173.2 XM_006722174.2 XM_006722175.2 XM_006722176.2 XM_006722177.3 XM_006722178.3 XM_011525471.2 XM_011525472.1 XM_017025326.1 XM_017025327.1 XM_017025328.1 XM_017025329.1 XM_017025330.1 XM_017025331.1 XM_017025332.1 XM_017025333.1 XM_017025334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7289803 c.5213G>A p.R1738H Chr.17: 58306353-58306353 on Build GRCh38
4498383 c.5197C>T p.P1733S Chr.17: 58306369-58306369 on Build GRCh38
4498459 c.5208C>T p.P1736P Chr.17: 58306358-58306358 on Build GRCh38
4498460 c.5208C>T p.P1736P Chr.17: 58306358-58306358 on Build GRCh38
3717491 c.5181C>A p.A1727A Chr.17: 58306385-58306385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762285520 Chr.17: 58306384-58306384 on Build GRCh38
rs191010956 Chr.17: 58306496-58306496 on Build GRCh38
rs760959614 Chr.17: 58306381-58306381 on Build GRCh38
rs201775210 Chr.17: 58306378-58306378 on Build GRCh38
rs199845425 Chr.17: 58306356-58306356 on Build GRCh38
rs563617114 Chr.17: 58306495-58306495 on Build GRCh38
rs752250359 Chr.17: 58306416-58306416 on Build GRCh38
rs915366488 Chr.17: 58306398-58306398 on Build GRCh38
rs754595190 Chr.17: 58306298-58306298 on Build GRCh38
rs747850524 Chr.17: 58306320-58306320 on Build GRCh38
rs1015707651 Chr.17: 58306347-58306347 on Build GRCh38
rs746281584 Chr.17: 58306293-58306293 on Build GRCh38
rs1003445726 Chr.17: 58306280-58306280 on Build GRCh38
rs34184983 Chr.17: 58306340-58306341 on Build GRCh38
rs777392694 Chr.17: 58306437-58306437 on Build GRCh38
rs919315805 Chr.17: 58306379-58306379 on Build GRCh38
rs1042249946 Chr.17: 58306434-58306434 on Build GRCh38
rs11079346 Chr.17: 58306383-58306383 on Build GRCh38
rs141114805 Chr.17: 58306414-58306414 on Build GRCh38
rs750646049 Chr.17: 58306271-58306271 on Build GRCh38
rs377098418 Chr.17: 58306395-58306395 on Build GRCh38
rs771037535 Chr.17: 58306354-58306354 on Build GRCh38
rs755623995 Chr.17: 58306468-58306468 on Build GRCh38
rs374407599 Chr.17: 58306309-58306309 on Build GRCh38
rs373474865 Chr.17: 58306452-58306452 on Build GRCh38
rs899762694 Chr.17: 58306439-58306439 on Build GRCh38
rs764042127 Chr.17: 58306382-58306382 on Build GRCh38
rs772806875 Chr.17: 58306353-58306353 on Build GRCh38
rs72839985 Chr.17: 58306329-58306329 on Build GRCh38
rs765774012 Chr.17: 58306393-58306393 on Build GRCh38
rs1049171322 Chr.17: 58306392-58306392 on Build GRCh38
rs369806515 Chr.17: 58306284-58306284 on Build GRCh38
rs965629389 Chr.17: 58306350-58306350 on Build GRCh38
rs1022872166 Chr.17: 58306287-58306288 on Build GRCh38
rs767771188 Chr.17: 58306364-58306364 on Build GRCh38
rs748843840 Chr.17: 58306440-58306440 on Build GRCh38
rs894792186 Chr.17: 58306291-58306291 on Build GRCh38
rs758631762 Chr.17: 58306403-58306403 on Build GRCh38
rs1027375542 Chr.17: 58306294-58306294 on Build GRCh38
rs892160219 Chr.17: 58306483-58306483 on Build GRCh38
rs375943233 Chr.17: 58306317-58306317 on Build GRCh38
rs755810475 Chr.17: 58306430-58306430 on Build GRCh38
rs956473534 Chr.17: 58306283-58306283 on Build GRCh38
rs759474747 Chr.17: 58306361-58306361 on Build GRCh38

View Full Product Details