Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4737767-4738016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf71
Entrez Gene ID
146562
Gene Name
chromosome 16 open reading frame 71
Gene Aliases
-
UniGene
Hs.602738
Gene Chromosome Location
Chr.16: 4734288-4749396 on Build GRCh38
Transcripts
NM_139170.2 XM_005255144.3 XM_017022975.1 XM_017022976.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144552 c.176C>T p.T59I Chr.16: 4737846-4737846 on Build GRCh38
7248792 c.264G>C p.E88D Chr.16: 4737934-4737934 on Build GRCh38
5479124 c.258T>C p.A86A Chr.16: 4737928-4737928 on Build GRCh38
4667005 c.249C>T p.V83V Chr.16: 4737919-4737919 on Build GRCh38
4060872 c.233A>C p.K78T Chr.16: 4737903-4737903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029302834 Chr.16: 4737988-4737988 on Build GRCh38
rs763561255 Chr.16: 4737985-4737985 on Build GRCh38
rs572256864 Chr.16: 4737788-4737788 on Build GRCh38
rs780953584 Chr.16: 4737822-4737822 on Build GRCh38
rs751908450 Chr.16: 4737933-4737933 on Build GRCh38
rs777884279 Chr.16: 4737807-4737807 on Build GRCh38
rs748777268 Chr.16: 4737887-4737887 on Build GRCh38
rs859310 Chr.16: 4737986-4737986 on Build GRCh38
rs752801300 Chr.16: 4737801-4737801 on Build GRCh38
rs767265532 Chr.16: 4737837-4737837 on Build GRCh38
rs138905202 Chr.16: 4737893-4737893 on Build GRCh38
rs764317984 Chr.16: 4737908-4737908 on Build GRCh38
rs780545004 Chr.16: 4737875-4737875 on Build GRCh38
rs758960991 Chr.16: 4737927-4737927 on Build GRCh38
rs757717935 Chr.16: 4737859-4737859 on Build GRCh38
rs754358461 Chr.16: 4737809-4737809 on Build GRCh38
rs200542507 Chr.16: 4737831-4737831 on Build GRCh38
rs751367804 Chr.16: 4737789-4737789 on Build GRCh38
rs761919932 Chr.16: 4737977-4737977 on Build GRCh38
rs937244059 Chr.16: 4737852-4737852 on Build GRCh38
rs747772347 Chr.16: 4737826-4737826 on Build GRCh38
rs138017310 Chr.16: 4737802-4737802 on Build GRCh38
rs749252507 Chr.16: 4737833-4737833 on Build GRCh38
rs774308542 Chr.16: 4737892-4737892 on Build GRCh38
rs778378677 Chr.16: 4737951-4737951 on Build GRCh38
rs747868724 Chr.16: 4737881-4737881 on Build GRCh38
rs765201393 Chr.16: 4737920-4737920 on Build GRCh38
rs765725967 Chr.16: 4737867-4737867 on Build GRCh38
rs200644126 Chr.16: 4737850-4737850 on Build GRCh38
rs763521364 Chr.16: 4737898-4737900 on Build GRCh38
rs868319711 Chr.16: 4737939-4737939 on Build GRCh38
rs760872155 Chr.16: 4737844-4737844 on Build GRCh38
rs149398988 Chr.16: 4737899-4737899 on Build GRCh38
rs779148305 Chr.16: 4737811-4737811 on Build GRCh38
rs770837155 Chr.16: 4737835-4737835 on Build GRCh38
rs141702834 Chr.16: 4737828-4737828 on Build GRCh38
rs768444062 Chr.16: 4737820-4737820 on Build GRCh38
rs748857655 Chr.16: 4737944-4737944 on Build GRCh38
rs779836621 Chr.16: 4737963-4737963 on Build GRCh38
rs991444368 Chr.16: 4737856-4737856 on Build GRCh38
rs777156319 Chr.16: 4737886-4737886 on Build GRCh38
rs771927622 Chr.16: 4737959-4737959 on Build GRCh38
rs760888954 Chr.16: 4737901-4737901 on Build GRCh38
rs545544504 Chr.16: 4737971-4737971 on Build GRCh38
rs527932688 Chr.16: 4737980-4737980 on Build GRCh38
rs746176285 Chr.16: 4737812-4737812 on Build GRCh38
rs199745071 Chr.16: 4737943-4737943 on Build GRCh38
rs17137215 Chr.16: 4737934-4737934 on Build GRCh38
rs200905968 Chr.16: 4737792-4737792 on Build GRCh38
rs867982424 Chr.16: 4737936-4737936 on Build GRCh38
rs759762627 Chr.16: 4737790-4737790 on Build GRCh38
rs74507482 Chr.16: 4737972-4737972 on Build GRCh38
rs755789871 Chr.16: 4737884-4737884 on Build GRCh38
rs745869622 Chr.16: 4737954-4737954 on Build GRCh38
rs150534835 Chr.16: 4737836-4737836 on Build GRCh38
rs766918761 Chr.16: 4737932-4737932 on Build GRCh38
rs775735405 Chr.16: 4737840-4737840 on Build GRCh38
rs893682173 Chr.16: 4737950-4737950 on Build GRCh38
rs115940628 Chr.16: 4737982-4737982 on Build GRCh38
rs770799787 Chr.16: 4737889-4737889 on Build GRCh38
rs757777580 Chr.16: 4737810-4737810 on Build GRCh38
rs146398065 Chr.16: 4737916-4737916 on Build GRCh38
rs146892525 Chr.16: 4737874-4737874 on Build GRCh38
rs1032551765 Chr.16: 4737795-4737795 on Build GRCh38
rs746769628 Chr.16: 4737966-4737966 on Build GRCh38
rs371625606 Chr.16: 4737847-4737847 on Build GRCh38
rs750849202 Chr.16: 4737868-4737868 on Build GRCh38
rs139743701 Chr.16: 4737919-4737919 on Build GRCh38

View Full Product Details