Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1322790-1323021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPTM1L
Entrez Gene ID
81037
Gene Name
CLPTM1 like
Gene Aliases
CRR9
UniGene
Hs.444673
Gene Chromosome Location
Chr.5: 1317744-1345070 on Build GRCh38
Transcripts
NM_030782.4 XM_011514144.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6168655 c.1289C>A p.S430Y Chr.5: 1322903-1322903 on Build GRCh38
1495912 c.1291T>C p.W431R Chr.5: 1322901-1322901 on Build GRCh38
4628360 c.1300A>G p.N434D Chr.5: 1322892-1322892 on Build GRCh38
3137735 c.1308C>T p.F436F Chr.5: 1322884-1322884 on Build GRCh38
4609114 c.1281-3C>A p.? Chr.5: 1322914-1322914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375686121 Chr.5: 1322823-1322823 on Build GRCh38
rs771816572 Chr.5: 1322851-1322851 on Build GRCh38
rs200308395 Chr.5: 1322839-1322839 on Build GRCh38
rs372360011 Chr.5: 1322918-1322918 on Build GRCh38
rs778591040 Chr.5: 1322827-1322827 on Build GRCh38
rs201228545 Chr.5: 1322883-1322883 on Build GRCh38
rs118151126 Chr.5: 1322987-1322987 on Build GRCh38
rs747797500 Chr.5: 1322928-1322930 on Build GRCh38
rs779529948 Chr.5: 1322921-1322921 on Build GRCh38
rs918765999 Chr.5: 1322845-1322845 on Build GRCh38
rs745697735 Chr.5: 1322937-1322937 on Build GRCh38
rs779794153 Chr.5: 1322956-1322956 on Build GRCh38
rs202096751 Chr.5: 1322884-1322884 on Build GRCh38
rs374530995 Chr.5: 1322874-1322874 on Build GRCh38
rs115028409 Chr.5: 1322878-1322878 on Build GRCh38
rs1041212936 Chr.5: 1322985-1322988 on Build GRCh38
rs374156376 Chr.5: 1322947-1322947 on Build GRCh38
rs142887696 Chr.5: 1322919-1322919 on Build GRCh38
rs780714790 Chr.5: 1322846-1322846 on Build GRCh38
rs146343560 Chr.5: 1322879-1322879 on Build GRCh38
rs946884345 Chr.5: 1322894-1322894 on Build GRCh38
rs374768502 Chr.5: 1322929-1322929 on Build GRCh38
rs367732058 Chr.5: 1322944-1322944 on Build GRCh38
rs753672415 Chr.5: 1322930-1322930 on Build GRCh38
rs982088160 Chr.5: 1322811-1322811 on Build GRCh38
rs772550989 Chr.5: 1322929-1322930 on Build GRCh38
rs748444750 Chr.5: 1322858-1322858 on Build GRCh38
rs371145678 Chr.5: 1322855-1322855 on Build GRCh38
rs201366704 Chr.5: 1322920-1322921 on Build GRCh38
rs752195731 Chr.5: 1322918-1322919 on Build GRCh38
rs746677864 Chr.5: 1322957-1322957 on Build GRCh38
rs889786197 Chr.5: 1322961-1322961 on Build GRCh38
rs577858144 Chr.5: 1322873-1322873 on Build GRCh38
rs959649220 Chr.5: 1322877-1322877 on Build GRCh38
rs996927211 Chr.5: 1322991-1322991 on Build GRCh38
rs756684328 Chr.5: 1322826-1322826 on Build GRCh38

View Full Product Details