Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 76192071-76192296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM266
Entrez Gene ID
123591
Gene Name
transmembrane protein 266
Gene Aliases
C15orf27, HVRP1
UniGene
Hs.661411
Gene Chromosome Location
Chr.15: 76059929-76204963 on Build GRCh38
Transcripts
NM_152335.2 XM_005254160.2 XM_017021915.1 XM_017021916.1 XM_017021917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5499222 c.953C>T p.S318L Chr.15: 76192152-76192152 on Build GRCh38
1198135 c.925G>A p.E309K Chr.15: 76192124-76192124 on Build GRCh38
4652637 c.931G>A p.V311I Chr.15: 76192130-76192130 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760576380 Chr.15: 76192119-76192119 on Build GRCh38
rs528967380 Chr.15: 76192207-76192207 on Build GRCh38
rs972717770 Chr.15: 76192227-76192227 on Build GRCh38
rs774320615 Chr.15: 76192135-76192135 on Build GRCh38
rs190417970 Chr.15: 76192117-76192117 on Build GRCh38
rs186450715 Chr.15: 76192101-76192101 on Build GRCh38
rs961569978 Chr.15: 76192223-76192223 on Build GRCh38
rs1045509885 Chr.15: 76192252-76192252 on Build GRCh38
rs907094360 Chr.15: 76192129-76192129 on Build GRCh38
rs986900388 Chr.15: 76192186-76192186 on Build GRCh38
rs574065572 Chr.15: 76192182-76192182 on Build GRCh38
rs949315988 Chr.15: 76192249-76192250 on Build GRCh38
rs919915458 Chr.15: 76192228-76192228 on Build GRCh38
rs1031306240 Chr.15: 76192173-76192173 on Build GRCh38
rs910885558 Chr.15: 76192187-76192187 on Build GRCh38
rs957176330 Chr.15: 76192178-76192178 on Build GRCh38
rs112753694 Chr.15: 76192218-76192218 on Build GRCh38
rs919764167 Chr.15: 76192211-76192211 on Build GRCh38
rs868440430 Chr.15: 76192266-76192266 on Build GRCh38
rs560224495 Chr.15: 76192202-76192202 on Build GRCh38
rs1012861884 Chr.15: 76192122-76192122 on Build GRCh38
rs141566322 Chr.15: 76192123-76192123 on Build GRCh38
rs11631826 Chr.15: 76192210-76192210 on Build GRCh38
rs931454790 Chr.15: 76192262-76192262 on Build GRCh38
rs552564746 Chr.15: 76192179-76192179 on Build GRCh38
rs905184288 Chr.15: 76192265-76192265 on Build GRCh38
rs894298372 Chr.15: 76192120-76192120 on Build GRCh38
rs534350468 Chr.15: 76192157-76192157 on Build GRCh38
rs917858496 Chr.15: 76192248-76192248 on Build GRCh38
rs1028515048 Chr.15: 76192201-76192201 on Build GRCh38

View Full Product Details