Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118998916-118999189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC84
Entrez Gene ID
338657
Gene Name
coiled-coil domain containing 84
Gene Aliases
DLNB14
UniGene
Hs.534613
Gene Chromosome Location
Chr.11: 118998067-119015793 on Build GRCh38
Transcripts
NM_198489.2 NR_104049.1 NR_104050.1 NR_104051.1 XM_011542799.1 XM_011542800.2 XM_017017652.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6220682 c.380C>A p.A127E Chr.11: 118999106-118999106 on Build GRCh38
4895188 c.346A>T p.K116* Chr.11: 118999072-118999072 on Build GRCh38
923691 c.351G>T p.E117D Chr.11: 118999077-118999077 on Build GRCh38
428527 c.371A>G p.Q124R Chr.11: 118999097-118999097 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782391125 Chr.11: 118998996-118998996 on Build GRCh38
rs782028375 Chr.11: 118999153-118999153 on Build GRCh38
rs781829160 Chr.11: 118999089-118999089 on Build GRCh38
rs782447292 Chr.11: 118999087-118999087 on Build GRCh38
rs782303233 Chr.11: 118999152-118999152 on Build GRCh38
rs1006668326 Chr.11: 118999047-118999047 on Build GRCh38
rs938747727 Chr.11: 118999107-118999107 on Build GRCh38
rs141547874 Chr.11: 118999078-118999078 on Build GRCh38
rs370163850 Chr.11: 118999120-118999120 on Build GRCh38
rs373706643 Chr.11: 118999143-118999143 on Build GRCh38
rs782756241 Chr.11: 118999083-118999083 on Build GRCh38
rs993653625 Chr.11: 118999148-118999148 on Build GRCh38
rs782679749 Chr.11: 118998988-118998988 on Build GRCh38
rs782816552 Chr.11: 118999041-118999041 on Build GRCh38
rs565442208 Chr.11: 118998939-118998939 on Build GRCh38
rs977907421 Chr.11: 118999057-118999057 on Build GRCh38
rs758493728 Chr.11: 118999134-118999134 on Build GRCh38
rs782546629 Chr.11: 118999108-118999108 on Build GRCh38
rs899394035 Chr.11: 118999135-118999135 on Build GRCh38
rs568193213 Chr.11: 118998943-118998943 on Build GRCh38
rs781909196 Chr.11: 118999118-118999118 on Build GRCh38
rs537730013 Chr.11: 118999116-118999116 on Build GRCh38
rs782126485 Chr.11: 118999073-118999073 on Build GRCh38
rs781982834 Chr.11: 118999003-118999003 on Build GRCh38
rs782100370 Chr.11: 118999022-118999022 on Build GRCh38
rs781938150 Chr.11: 118999034-118999034 on Build GRCh38
rs375803206 Chr.11: 118999108-118999108 on Build GRCh38
rs782284632 Chr.11: 118998993-118998993 on Build GRCh38
rs140617350 Chr.11: 118999031-118999031 on Build GRCh38
rs782617592 Chr.11: 118998978-118998978 on Build GRCh38
rs1013312169 Chr.11: 118999139-118999139 on Build GRCh38
rs782197824 Chr.11: 118999149-118999149 on Build GRCh38
rs782590650 Chr.11: 118999114-118999114 on Build GRCh38
rs373436467 Chr.11: 118999035-118999035 on Build GRCh38
rs782246995 Chr.11: 118999132-118999132 on Build GRCh38
rs782498387 Chr.11: 118998977-118998977 on Build GRCh38
rs782198970 Chr.11: 118998982-118998982 on Build GRCh38
rs61902079 Chr.11: 118999123-118999123 on Build GRCh38
rs782316318 Chr.11: 118998986-118998986 on Build GRCh38

View Full Product Details