Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108648424-108648695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1C
Entrez Gene ID
23603
Gene Name
coronin 1C
Gene Aliases
HCRNN4
UniGene
Hs.330384
Gene Chromosome Location
Chr.12: 108645109-108731550 on Build GRCh38
Transcripts
NM_001105237.2 NM_001276471.1 NM_014325.3 XM_011538125.1 XM_017019121.1 XM_017019122.1 XM_017019123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634228 c.1420G>A p.D474N Chr.12: 108648649-108648649 on Build GRCh38
5962782 c.1282A>T p.K428* Chr.12: 108648628-108648628 on Build GRCh38
6520644 c.1239G>T p.K413N Chr.12: 108648671-108648671 on Build GRCh38
3737492 c.1264C>A p.L422M Chr.12: 108648646-108648646 on Build GRCh38
6520645 c.1398G>T p.K466N Chr.12: 108648671-108648671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775054781 Chr.12: 108648641-108648641 on Build GRCh38
rs752931977 Chr.12: 108648528-108648528 on Build GRCh38
rs894176192 Chr.12: 108648510-108648510 on Build GRCh38
rs139386424 Chr.12: 108648607-108648607 on Build GRCh38
rs761544630 Chr.12: 108648661-108648661 on Build GRCh38
rs939101553 Chr.12: 108648623-108648623 on Build GRCh38
rs765047650 Chr.12: 108648666-108648666 on Build GRCh38
rs535970064 Chr.12: 108648614-108648614 on Build GRCh38
rs1043995474 Chr.12: 108648455-108648455 on Build GRCh38
rs375125043 Chr.12: 108648590-108648590 on Build GRCh38
rs913606663 Chr.12: 108648663-108648663 on Build GRCh38
rs778431340 Chr.12: 108648609-108648609 on Build GRCh38
rs542158409 Chr.12: 108648503-108648503 on Build GRCh38
rs139820945 Chr.12: 108648578-108648578 on Build GRCh38
rs754972843 Chr.12: 108648585-108648585 on Build GRCh38
rs950712241 Chr.12: 108648559-108648559 on Build GRCh38
rs1017741365 Chr.12: 108648523-108648523 on Build GRCh38
rs571036715 Chr.12: 108648543-108648543 on Build GRCh38
rs201118257 Chr.12: 108648646-108648646 on Build GRCh38
rs757195237 Chr.12: 108648669-108648669 on Build GRCh38
rs552641527 Chr.12: 108648542-108648542 on Build GRCh38
rs77747402 Chr.12: 108648548-108648548 on Build GRCh38
rs79808687 Chr.12: 108648526-108648526 on Build GRCh38
rs991919716 Chr.12: 108648640-108648640 on Build GRCh38
rs531228040 Chr.12: 108648535-108648535 on Build GRCh38
rs368475426 Chr.12: 108648615-108648615 on Build GRCh38
rs746564481 Chr.12: 108648644-108648644 on Build GRCh38
rs191109349 Chr.12: 108648633-108648633 on Build GRCh38
rs928545705 Chr.12: 108648469-108648469 on Build GRCh38
rs368755225 Chr.12: 108648572-108648572 on Build GRCh38
rs962633522 Chr.12: 108648612-108648612 on Build GRCh38
rs745506701 Chr.12: 108648621-108648621 on Build GRCh38
rs749607320 Chr.12: 108648610-108648610 on Build GRCh38
rs776558672 Chr.12: 108648658-108648658 on Build GRCh38
rs1055402268 Chr.12: 108648504-108648504 on Build GRCh38
rs761596232 Chr.12: 108648566-108648566 on Build GRCh38
rs772919938 Chr.12: 108648668-108648668 on Build GRCh38
rs768511383 Chr.12: 108648649-108648649 on Build GRCh38
rs904052741 Chr.12: 108648479-108648479 on Build GRCh38

View Full Product Details