Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6253683-6253948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR153
Entrez Gene ID
387509
Gene Name
G protein-coupled receptor 153
Gene Aliases
PGR1
UniGene
Hs.531581
Gene Chromosome Location
Chr.1: 6247346-6260975 on Build GRCh38
Transcripts
NM_207370.2 XM_011541434.2 XM_017001250.1 XM_017001251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5977822 c.677G>A p.R226Q Chr.1: 6253827-6253827 on Build GRCh38
325422 c.601G>A p.V201M Chr.1: 6253903-6253903 on Build GRCh38
4009280 c.723G>A p.Q241Q Chr.1: 6253781-6253781 on Build GRCh38
7131639 c.716C>T p.S239F Chr.1: 6253788-6253788 on Build GRCh38
3386280 c.760G>A p.D254N Chr.1: 6253744-6253744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961328298 Chr.1: 6253768-6253768 on Build GRCh38
rs776072994 Chr.1: 6253706-6253706 on Build GRCh38
rs777097027 Chr.1: 6253874-6253874 on Build GRCh38
rs570252149 Chr.1: 6253921-6253921 on Build GRCh38
rs778611348 Chr.1: 6253816-6253816 on Build GRCh38
rs754228590 Chr.1: 6253860-6253860 on Build GRCh38
rs12735670 Chr.1: 6253878-6253878 on Build GRCh38
rs147103865 Chr.1: 6253775-6253775 on Build GRCh38
rs375133105 Chr.1: 6253709-6253709 on Build GRCh38
rs766020613 Chr.1: 6253776-6253776 on Build GRCh38
rs756518447 Chr.1: 6253810-6253810 on Build GRCh38
rs751705076 Chr.1: 6253833-6253833 on Build GRCh38
rs149182010 Chr.1: 6253824-6253824 on Build GRCh38
rs781477203 Chr.1: 6253805-6253805 on Build GRCh38
rs761003756 Chr.1: 6253897-6253897 on Build GRCh38
rs755398589 Chr.1: 6253906-6253906 on Build GRCh38
rs762831977 Chr.1: 6253888-6253888 on Build GRCh38
rs566005645 Chr.1: 6253883-6253883 on Build GRCh38
rs757383254 Chr.1: 6253769-6253769 on Build GRCh38
rs764491787 Chr.1: 6253868-6253870 on Build GRCh38
rs776469038 Chr.1: 6253877-6253877 on Build GRCh38
rs753825747 Chr.1: 6253781-6253781 on Build GRCh38
rs61739500 Chr.1: 6253924-6253924 on Build GRCh38
rs146978653 Chr.1: 6253882-6253883 on Build GRCh38
rs755596313 Chr.1: 6253861-6253861 on Build GRCh38
rs375868582 Chr.1: 6253875-6253875 on Build GRCh38
rs761890147 Chr.1: 6253838-6253838 on Build GRCh38
rs571664815 Chr.1: 6253841-6253841 on Build GRCh38
rs377668413 Chr.1: 6253744-6253744 on Build GRCh38
rs139578429 Chr.1: 6253865-6253865 on Build GRCh38
rs897234991 Chr.1: 6253889-6253889 on Build GRCh38
rs759431353 Chr.1: 6253789-6253789 on Build GRCh38
rs772922075 Chr.1: 6253822-6253822 on Build GRCh38
rs767251936 Chr.1: 6253749-6253749 on Build GRCh38
rs769251010 Chr.1: 6253814-6253814 on Build GRCh38
rs189356842 Chr.1: 6253798-6253798 on Build GRCh38
rs144648218 Chr.1: 6253745-6253745 on Build GRCh38
rs758789216 Chr.1: 6253912-6253912 on Build GRCh38
rs111809364 Chr.1: 6253914-6253914 on Build GRCh38
rs749841451 Chr.1: 6253813-6253813 on Build GRCh38
rs748238110 Chr.1: 6253911-6253911 on Build GRCh38
rs143332760 Chr.1: 6253827-6253827 on Build GRCh38
rs888633434 Chr.1: 6253735-6253735 on Build GRCh38
rs770274504 Chr.1: 6253705-6253705 on Build GRCh38
rs780504968 Chr.1: 6253852-6253852 on Build GRCh38
rs770779811 Chr.1: 6253825-6253825 on Build GRCh38
rs772938106 Chr.1: 6253756-6253756 on Build GRCh38
rs757002132 Chr.1: 6253927-6253927 on Build GRCh38
rs144536689 Chr.1: 6253903-6253903 on Build GRCh38
rs767669310 Chr.1: 6253890-6253890 on Build GRCh38
rs757533205 Chr.1: 6253799-6253799 on Build GRCh38
rs752052507 Chr.1: 6253876-6253876 on Build GRCh38
rs750547357 Chr.1: 6253806-6253806 on Build GRCh38
rs137859776 Chr.1: 6253862-6253862 on Build GRCh38
rs200131017 Chr.1: 6253891-6253891 on Build GRCh38
rs747644048 Chr.1: 6253818-6253818 on Build GRCh38
rs746234507 Chr.1: 6253702-6253702 on Build GRCh38
rs1035922375 Chr.1: 6253864-6253864 on Build GRCh38
rs765639331 Chr.1: 6253910-6253910 on Build GRCh38
rs370700931 Chr.1: 6253708-6253708 on Build GRCh38
rs769515193 Chr.1: 6253879-6253879 on Build GRCh38
rs767440916 Chr.1: 6253839-6253839 on Build GRCh38
rs141201270 Chr.1: 6253840-6253840 on Build GRCh38
rs531536988 Chr.1: 6253828-6253828 on Build GRCh38
rs537787050 Chr.1: 6253925-6253925 on Build GRCh38
rs771623780 Chr.1: 6253873-6253873 on Build GRCh38
rs781180390 Chr.1: 6253904-6253904 on Build GRCh38
rs765147731 Chr.1: 6253829-6253829 on Build GRCh38
rs369597145 Chr.1: 6253887-6253887 on Build GRCh38
rs982255285 Chr.1: 6253855-6253857 on Build GRCh38
rs765766507 Chr.1: 6253820-6253820 on Build GRCh38
rs771688682 Chr.1: 6253819-6253819 on Build GRCh38
rs186481236 Chr.1: 6253882-6253882 on Build GRCh38

View Full Product Details