Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186571924-186572173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB11
Entrez Gene ID
51726
Gene Name
DnaJ heat shock protein family (Hsp40) member B11
Gene Aliases
ABBP-2, ABBP2, DJ9, Dj-9, EDJ, ERdj3, ERj3, ERj3p, PRO1080, UNQ537
UniGene
Hs.317192
Gene Chromosome Location
Chr.3: 186570676-186585800 on Build GRCh38
Transcripts
NM_016306.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6286153 c.73G>T p.D25Y Chr.3: 186572099-186572099 on Build GRCh38
5355394 c.104G>A p.S35N Chr.3: 186572130-186572130 on Build GRCh38
4625040 c.71G>A p.R24Q Chr.3: 186572097-186572097 on Build GRCh38
1421493 c.70C>T p.R24* Chr.3: 186572096-186572096 on Build GRCh38
1042310 c.76T>G p.F26V Chr.3: 186572102-186572102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761209634 Chr.3: 186572088-186572088 on Build GRCh38
rs3832261 Chr.3: 186572081-186572084 on Build GRCh38
rs767931261 Chr.3: 186572140-186572140 on Build GRCh38
rs562998711 Chr.3: 186571959-186571959 on Build GRCh38
rs778052998 Chr.3: 186572062-186572062 on Build GRCh38
rs764560443 Chr.3: 186572095-186572095 on Build GRCh38
rs972021081 Chr.3: 186572122-186572122 on Build GRCh38
rs751942743 Chr.3: 186572064-186572064 on Build GRCh38
rs952060662 Chr.3: 186572143-186572143 on Build GRCh38
rs1010768921 Chr.3: 186572061-186572061 on Build GRCh38
rs755066814 Chr.3: 186572078-186572081 on Build GRCh38
rs138527662 Chr.3: 186571987-186571987 on Build GRCh38
rs1034241493 Chr.3: 186572071-186572071 on Build GRCh38
rs745441260 Chr.3: 186571981-186571981 on Build GRCh38
rs372242752 Chr.3: 186572065-186572065 on Build GRCh38
rs750148501 Chr.3: 186572138-186572138 on Build GRCh38
rs1046123787 Chr.3: 186571968-186571968 on Build GRCh38
rs775825031 Chr.3: 186572082-186572082 on Build GRCh38
rs934962950 Chr.3: 186572025-186572025 on Build GRCh38
rs763526735 Chr.3: 186571971-186571971 on Build GRCh38
rs774633788 Chr.3: 186572066-186572066 on Build GRCh38
rs768512470 Chr.3: 186572035-186572035 on Build GRCh38
rs148976481 Chr.3: 186572126-186572126 on Build GRCh38
rs371162691 Chr.3: 186572060-186572060 on Build GRCh38
rs199514974 Chr.3: 186572137-186572137 on Build GRCh38
rs1052051221 Chr.3: 186572031-186572031 on Build GRCh38
rs375290424 Chr.3: 186572078-186572078 on Build GRCh38
rs893511129 Chr.3: 186572051-186572051 on Build GRCh38
rs757507181 Chr.3: 186572080-186572080 on Build GRCh38
rs777074056 Chr.3: 186572106-186572106 on Build GRCh38
rs904462679 Chr.3: 186572019-186572019 on Build GRCh38
rs1014467656 Chr.3: 186572086-186572086 on Build GRCh38
rs200721132 Chr.3: 186572070-186572070 on Build GRCh38
rs894334420 Chr.3: 186572073-186572073 on Build GRCh38

View Full Product Details