Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42962949-42963186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY3-GNMT
Entrez Gene ID
107080644
Gene Name
CNPY3-GNMT readthrough
Gene Aliases
-
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42929130-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Gene Symbol
GNMT
Entrez Gene ID
27232
Gene Name
glycine N-methyltransferase
Gene Aliases
HEL-S-182mP
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42960754-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6513967 c.478C>A p.L160M Chr.6: 42963098-42963098 on Build GRCh38
4687498 c.554G>A p.S185N Chr.6: 42963174-42963174 on Build GRCh38
3874152 c.491C>T p.A164V Chr.6: 42963111-42963111 on Build GRCh38
1621669 c.541G>A p.D181N Chr.6: 42963161-42963161 on Build GRCh38
3874151 c.466C>T p.H156Y Chr.6: 42963086-42963086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146275920 Chr.6: 42963074-42963074 on Build GRCh38
rs138085632 Chr.6: 42963087-42963087 on Build GRCh38
rs201556777 Chr.6: 42963030-42963030 on Build GRCh38
rs121907889 Chr.6: 42963149-42963149 on Build GRCh38
rs781476590 Chr.6: 42963096-42963096 on Build GRCh38
rs1031947375 Chr.6: 42962989-42962989 on Build GRCh38
rs149792195 Chr.6: 42963112-42963112 on Build GRCh38
rs147291064 Chr.6: 42963139-42963139 on Build GRCh38
rs752057158 Chr.6: 42963142-42963142 on Build GRCh38
rs144716335 Chr.6: 42963123-42963123 on Build GRCh38
rs371794899 Chr.6: 42963130-42963130 on Build GRCh38
rs200015105 Chr.6: 42963057-42963057 on Build GRCh38
rs577108844 Chr.6: 42962976-42962976 on Build GRCh38
rs780155247 Chr.6: 42963107-42963107 on Build GRCh38
rs750649643 Chr.6: 42963097-42963097 on Build GRCh38
rs147746130 Chr.6: 42963160-42963160 on Build GRCh38
rs762369159 Chr.6: 42963144-42963144 on Build GRCh38
rs770519701 Chr.6: 42963128-42963128 on Build GRCh38
rs200543916 Chr.6: 42963165-42963165 on Build GRCh38
rs778228488 Chr.6: 42963126-42963136 on Build GRCh38
rs200406766 Chr.6: 42963056-42963056 on Build GRCh38
rs1014621237 Chr.6: 42963046-42963046 on Build GRCh38
rs772909903 Chr.6: 42963058-42963058 on Build GRCh38
rs771883382 Chr.6: 42963121-42963121 on Build GRCh38
rs566244116 Chr.6: 42963153-42963153 on Build GRCh38
rs202053748 Chr.6: 42963079-42963079 on Build GRCh38
rs1000528575 Chr.6: 42963027-42963027 on Build GRCh38
rs779187760 Chr.6: 42963163-42963163 on Build GRCh38
rs142613217 Chr.6: 42963089-42963089 on Build GRCh38
rs998590857 Chr.6: 42962984-42962984 on Build GRCh38
rs1012093959 Chr.6: 42963039-42963039 on Build GRCh38
rs145997766 Chr.6: 42963108-42963108 on Build GRCh38
rs201256313 Chr.6: 42963150-42963150 on Build GRCh38
rs904285290 Chr.6: 42962981-42962981 on Build GRCh38
rs563826316 Chr.6: 42963090-42963090 on Build GRCh38
rs756343530 Chr.6: 42963102-42963102 on Build GRCh38
rs772929804 Chr.6: 42963122-42963122 on Build GRCh38
rs749218665 Chr.6: 42963029-42963029 on Build GRCh38
rs768654043 Chr.6: 42963111-42963111 on Build GRCh38
rs1042140749 Chr.6: 42962986-42962986 on Build GRCh38
rs768087069 Chr.6: 42963088-42963088 on Build GRCh38
rs755100354 Chr.6: 42963161-42963161 on Build GRCh38
rs759061456 Chr.6: 42963132-42963132 on Build GRCh38
rs753327439 Chr.6: 42963080-42963080 on Build GRCh38
rs769917180 Chr.6: 42963025-42963025 on Build GRCh38
rs1022889192 Chr.6: 42963052-42963052 on Build GRCh38
rs774236296 Chr.6: 42963047-42963047 on Build GRCh38

View Full Product Details