Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26634743-26635000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL2
Entrez Gene ID
1808
Gene Name
dihydropyrimidinase like 2
Gene Aliases
CRMP-2, CRMP2, DHPRP2, DRP-2, DRP2, N2A3, ULIP-2, ULIP2
UniGene
Hs.593187
Gene Chromosome Location
Chr.8: 26514193-26658177 on Build GRCh38
Transcripts
NM_001197293.2 NM_001244604.1 NM_001386.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1098544 c.799G>A p.A267T Chr.8: 26634888-26634888 on Build GRCh38
300049 c.727G>A p.A243T Chr.8: 26634816-26634816 on Build GRCh38
1456304 c.803G>A p.R268Q Chr.8: 26634892-26634892 on Build GRCh38
1098543 c.771C>T p.S257S Chr.8: 26634860-26634860 on Build GRCh38
1098542 c.712C>T p.R238C Chr.8: 26634801-26634801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775541793 Chr.8: 26634918-26634918 on Build GRCh38
rs758968723 Chr.8: 26634879-26634879 on Build GRCh38
rs950954006 Chr.8: 26634953-26634953 on Build GRCh38
rs113429984 Chr.8: 26634853-26634853 on Build GRCh38
rs1016382767 Chr.8: 26634966-26634966 on Build GRCh38
rs542156396 Chr.8: 26634964-26634964 on Build GRCh38
rs540612356 Chr.8: 26634807-26634807 on Build GRCh38
rs569990259 Chr.8: 26634782-26634782 on Build GRCh38
rs970694512 Chr.8: 26634805-26634805 on Build GRCh38
rs761564575 Chr.8: 26634942-26634942 on Build GRCh38
rs113106776 Chr.8: 26634923-26634923 on Build GRCh38
rs750894336 Chr.8: 26634872-26634872 on Build GRCh38
rs768235949 Chr.8: 26634940-26634940 on Build GRCh38
rs529587582 Chr.8: 26634788-26634788 on Build GRCh38
rs78121726 Chr.8: 26634881-26634881 on Build GRCh38
rs944330260 Chr.8: 26634889-26634889 on Build GRCh38
rs748387219 Chr.8: 26634895-26634895 on Build GRCh38
rs576395295 Chr.8: 26634956-26634956 on Build GRCh38
rs141745272 Chr.8: 26634836-26634836 on Build GRCh38
rs747179035 Chr.8: 26634774-26634774 on Build GRCh38
rs758496602 Chr.8: 26634766-26634766 on Build GRCh38
rs765712904 Chr.8: 26634869-26634869 on Build GRCh38
rs778681165 Chr.8: 26634909-26634909 on Build GRCh38
rs35690946 Chr.8: 26634958-26634959 on Build GRCh38
rs768747449 Chr.8: 26634777-26634777 on Build GRCh38
rs751569597 Chr.8: 26634882-26634882 on Build GRCh38
rs770652042 Chr.8: 26634901-26634901 on Build GRCh38
rs565567744 Chr.8: 26634829-26634829 on Build GRCh38
rs959112805 Chr.8: 26634837-26634837 on Build GRCh38
rs539422560 Chr.8: 26634941-26634941 on Build GRCh38
rs921303034 Chr.8: 26634903-26634903 on Build GRCh38
rs770739859 Chr.8: 26634794-26634794 on Build GRCh38
rs1041057063 Chr.8: 26634899-26634899 on Build GRCh38
rs147069060 Chr.8: 26634839-26634839 on Build GRCh38
rs148064770 Chr.8: 26634888-26634888 on Build GRCh38
rs746399465 Chr.8: 26634764-26634764 on Build GRCh38
rs774852669 Chr.8: 26634826-26634826 on Build GRCh38
rs760066736 Chr.8: 26634933-26634933 on Build GRCh38
rs112794605 Chr.8: 26634781-26634781 on Build GRCh38
rs55906521 Chr.8: 26634954-26634954 on Build GRCh38
rs35621323 Chr.8: 26634815-26634815 on Build GRCh38
rs376515723 Chr.8: 26634772-26634772 on Build GRCh38
rs111498189 Chr.8: 26634979-26634979 on Build GRCh38
rs1032288349 Chr.8: 26634776-26634776 on Build GRCh38
rs920061983 Chr.8: 26634862-26634862 on Build GRCh38
rs745724140 Chr.8: 26634915-26634915 on Build GRCh38
rs373662465 Chr.8: 26634820-26634820 on Build GRCh38

View Full Product Details