Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64232337-64232588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC4
Entrez Gene ID
3338
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
Gene Aliases
DANJC4, HSPF2, MCG18
UniGene
Hs.172847
Gene Chromosome Location
Chr.11: 64230281-64234281 on Build GRCh38
Transcripts
NM_001307980.1 NM_001307981.1 NM_005528.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400822 c.310C>T p.P104S Chr.11: 64232559-64232559 on Build GRCh38
4146047 c.281G>C p.S94T Chr.11: 64232530-64232530 on Build GRCh38
689746 c.91C>A p.L31M Chr.11: 64232463-64232463 on Build GRCh38
7321282 c.241G>A p.E81K Chr.11: 64232490-64232490 on Build GRCh38
4035071 c.222C>T p.S74S Chr.11: 64232471-64232471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764043069 Chr.11: 64232488-64232488 on Build GRCh38
rs774284402 Chr.11: 64232533-64232533 on Build GRCh38
rs571900284 Chr.11: 64232458-64232458 on Build GRCh38
rs979210242 Chr.11: 64232450-64232450 on Build GRCh38
rs749756355 Chr.11: 64232460-64232460 on Build GRCh38
rs756469089 Chr.11: 64232503-64232503 on Build GRCh38
rs200699204 Chr.11: 64232510-64232510 on Build GRCh38
rs369456737 Chr.11: 64232448-64232448 on Build GRCh38
rs199745306 Chr.11: 64232457-64232457 on Build GRCh38
rs775103818 Chr.11: 64232475-64232475 on Build GRCh38
rs760217798 Chr.11: 64232385-64232385 on Build GRCh38
rs761896083 Chr.11: 64232535-64232535 on Build GRCh38
rs750527086 Chr.11: 64232546-64232546 on Build GRCh38
rs757140841 Chr.11: 64232443-64232443 on Build GRCh38
rs879710727 Chr.11: 64232373-64232373 on Build GRCh38
rs763012788 Chr.11: 64232427-64232427 on Build GRCh38
rs117464526 Chr.11: 64232547-64232547 on Build GRCh38
rs569419878 Chr.11: 64232357-64232357 on Build GRCh38
rs764733432 Chr.11: 64232414-64232414 on Build GRCh38
rs769533900 Chr.11: 64232524-64232524 on Build GRCh38
rs769249584 Chr.11: 64232465-64232465 on Build GRCh38
rs370287965 Chr.11: 64232530-64232530 on Build GRCh38
rs745534264 Chr.11: 64232473-64232473 on Build GRCh38
rs766341609 Chr.11: 64232505-64232505 on Build GRCh38
rs759008176 Chr.11: 64232408-64232408 on Build GRCh38
rs781520025 Chr.11: 64232447-64232447 on Build GRCh38
rs778170358 Chr.11: 64232517-64232517 on Build GRCh38
rs950719391 Chr.11: 64232416-64232416 on Build GRCh38
rs200183856 Chr.11: 64232526-64232526 on Build GRCh38
rs750657148 Chr.11: 64232500-64232500 on Build GRCh38
rs752521946 Chr.11: 64232424-64232424 on Build GRCh38
rs140273970 Chr.11: 64232356-64232356 on Build GRCh38
rs201697121 Chr.11: 64232527-64232527 on Build GRCh38
rs751418913 Chr.11: 64232442-64232442 on Build GRCh38
rs776288235 Chr.11: 64232400-64232400 on Build GRCh38
rs748467006 Chr.11: 64232467-64232467 on Build GRCh38
rs528878768 Chr.11: 64232490-64232490 on Build GRCh38
rs201362496 Chr.11: 64232529-64232529 on Build GRCh38
rs776200378 Chr.11: 64232472-64232472 on Build GRCh38
rs370434882 Chr.11: 64232515-64232515 on Build GRCh38
rs541349768 Chr.11: 64232479-64232479 on Build GRCh38
rs767413232 Chr.11: 64232543-64232543 on Build GRCh38
rs201258154 Chr.11: 64232489-64232489 on Build GRCh38
rs193059245 Chr.11: 64232499-64232499 on Build GRCh38
rs143213764 Chr.11: 64232461-64232462 on Build GRCh38
rs879131289 Chr.11: 64232419-64232419 on Build GRCh38
rs899358431 Chr.11: 64232553-64232553 on Build GRCh38
rs562381353 Chr.11: 64232512-64232512 on Build GRCh38
rs760961158 Chr.11: 64232548-64232548 on Build GRCh38
rs377031581 Chr.11: 64232379-64232379 on Build GRCh38
rs766577923 Chr.11: 64232555-64232555 on Build GRCh38
rs767684425 Chr.11: 64232414-64232414 on Build GRCh38
rs986889001 Chr.11: 64232387-64232387 on Build GRCh38
rs770623397 Chr.11: 64232395-64232395 on Build GRCh38
rs772458068 Chr.11: 64232468-64232468 on Build GRCh38
rs919262581 Chr.11: 64232423-64232423 on Build GRCh38
rs369570434 Chr.11: 64232390-64232390 on Build GRCh38
rs374099725 Chr.11: 64232523-64232523 on Build GRCh38
rs779129165 Chr.11: 64232466-64232466 on Build GRCh38
rs375824405 Chr.11: 64232513-64232513 on Build GRCh38
rs763763877 Chr.11: 64232437-64232437 on Build GRCh38
rs755040167 Chr.11: 64232511-64232511 on Build GRCh38
rs766471862 Chr.11: 64232368-64232368 on Build GRCh38

View Full Product Details