Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233146264-233146511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5D
Entrez Gene ID
3635
Gene Name
inositol polyphosphate-5-phosphatase D
Gene Aliases
SHIP, SHIP-1, SHIP1, SIP-145, hp51CN, p150Ship
UniGene
Hs.262886
Gene Chromosome Location
Chr.2: 233059967-233207903 on Build GRCh38
Transcripts
NM_001017915.2 NM_005541.4 XM_017004004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5532744 c.883C>T p.L295F Chr.2: 233146415-233146415 on Build GRCh38
7262850 c.835-8G>A p.? Chr.2: 233146359-233146359 on Build GRCh38
7259882 c.874C>T p.R292W Chr.2: 233146406-233146406 on Build GRCh38
1614526 c.888C>T p.I296I Chr.2: 233146420-233146420 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749585042 Chr.2: 233146484-233146497 on Build GRCh38
rs930906898 Chr.2: 233146389-233146389 on Build GRCh38
rs573550640 Chr.2: 233146407-233146407 on Build GRCh38
rs574229969 Chr.2: 233146359-233146359 on Build GRCh38
rs190815070 Chr.2: 233146402-233146402 on Build GRCh38
rs887882626 Chr.2: 233146396-233146396 on Build GRCh38
rs531004050 Chr.2: 233146473-233146473 on Build GRCh38
rs768271677 Chr.2: 233146448-233146448 on Build GRCh38
rs781760700 Chr.2: 233146361-233146361 on Build GRCh38
rs752898160 Chr.2: 233146338-233146338 on Build GRCh38
rs565606610 Chr.2: 233146458-233146458 on Build GRCh38
rs571330134 Chr.2: 233146325-233146325 on Build GRCh38
rs36189208 Chr.2: 233146358-233146358 on Build GRCh38
rs747649456 Chr.2: 233146462-233146462 on Build GRCh38
rs1000562398 Chr.2: 233146307-233146307 on Build GRCh38
rs757810007 Chr.2: 233146401-233146401 on Build GRCh38
rs777385833 Chr.2: 233146480-233146480 on Build GRCh38
rs545697179 Chr.2: 233146411-233146411 on Build GRCh38
rs956097436 Chr.2: 233146328-233146328 on Build GRCh38
rs766135487 Chr.2: 233146327-233146327 on Build GRCh38
rs150609958 Chr.2: 233146474-233146474 on Build GRCh38
rs543005069 Chr.2: 233146384-233146384 on Build GRCh38
rs780143535 Chr.2: 233146385-233146385 on Build GRCh38
rs534617490 Chr.2: 233146298-233146298 on Build GRCh38
rs145465825 Chr.2: 233146324-233146324 on Build GRCh38
rs773888093 Chr.2: 233146460-233146460 on Build GRCh38
rs748910944 Chr.2: 233146424-233146424 on Build GRCh38
rs971008688 Chr.2: 233146353-233146353 on Build GRCh38
rs758598356 Chr.2: 233146339-233146339 on Build GRCh38
rs988504712 Chr.2: 233146342-233146342 on Build GRCh38
rs758641144 Chr.2: 233146362-233146362 on Build GRCh38
rs36180605 Chr.2: 233146343-233146343 on Build GRCh38

View Full Product Details