Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67181731-67181994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2A
Entrez Gene ID
22992
Gene Name
lysine demethylase 2A
Gene Aliases
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
UniGene
Hs.124147
Gene Chromosome Location
Chr.11: 67119210-67258082 on Build GRCh38
Transcripts
NM_012308.2 NR_027473.1 XM_011544860.2 XM_017017391.1 XM_017017392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1239207 c.267G>A p.P89P Chr.11: 67181852-67181852 on Build GRCh38
3383761 c.307+1G>T p.? Chr.11: 67181893-67181893 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759185120 Chr.11: 67181905-67181905 on Build GRCh38
rs368849359 Chr.11: 67181901-67181901 on Build GRCh38
rs372756373 Chr.11: 67181864-67181864 on Build GRCh38
rs776578589 Chr.11: 67181902-67181902 on Build GRCh38
rs1055521406 Chr.11: 67181946-67181946 on Build GRCh38
rs752036560 Chr.11: 67181805-67181805 on Build GRCh38
rs146139491 Chr.11: 67181803-67181803 on Build GRCh38
rs752242185 Chr.11: 67181910-67181910 on Build GRCh38
rs977249682 Chr.11: 67181913-67181913 on Build GRCh38
rs764972194 Chr.11: 67181906-67181906 on Build GRCh38
rs990577693 Chr.11: 67181879-67181879 on Build GRCh38
rs367881055 Chr.11: 67181831-67181831 on Build GRCh38
rs149640085 Chr.11: 67181802-67181802 on Build GRCh38
rs773324257 Chr.11: 67181867-67181867 on Build GRCh38
rs868120492 Chr.11: 67181850-67181850 on Build GRCh38
rs555338591 Chr.11: 67181843-67181843 on Build GRCh38
rs762568955 Chr.11: 67181933-67181933 on Build GRCh38
rs371911656 Chr.11: 67181963-67181963 on Build GRCh38
rs919580026 Chr.11: 67181760-67181760 on Build GRCh38
rs573341763 Chr.11: 67181944-67181944 on Build GRCh38
rs367910857 Chr.11: 67181796-67181796 on Build GRCh38
rs894107998 Chr.11: 67181781-67181781 on Build GRCh38
rs770062376 Chr.11: 67181834-67181834 on Build GRCh38
rs1015082729 Chr.11: 67181824-67181824 on Build GRCh38
rs757655739 Chr.11: 67181810-67181810 on Build GRCh38
rs754046694 Chr.11: 67181852-67181852 on Build GRCh38
rs780102851 Chr.11: 67181841-67181841 on Build GRCh38
rs749515965 Chr.11: 67181838-67181838 on Build GRCh38
rs931239200 Chr.11: 67181776-67181776 on Build GRCh38
rs760687327 Chr.11: 67181891-67181891 on Build GRCh38
rs768755482 Chr.11: 67181919-67181919 on Build GRCh38
rs755283965 Chr.11: 67181945-67181945 on Build GRCh38
rs781530899 Chr.11: 67181819-67181819 on Build GRCh38
rs375732510 Chr.11: 67181865-67181865 on Build GRCh38

View Full Product Details