Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93215326-93215587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEPACAM2
Entrez Gene ID
253012
Gene Name
HEPACAM family member 2
Gene Aliases
MIKI
UniGene
Hs.443169
Gene Chromosome Location
Chr.7: 93188586-93232297 on Build GRCh38
Transcripts
NM_001039372.2 NM_001288804.1 NM_001288810.1 NM_198151.2 XM_011516000.2 XM_011516001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4947726 c.714T>A p.Y238* Chr.7: 93215402-93215402 on Build GRCh38
1452926 c.554delA p.N185fs*25 Chr.7: 93215562-93215562 on Build GRCh38
5887983 c.644G>A p.G215E Chr.7: 93215472-93215472 on Build GRCh38
3924238 c.521G>A p.G174E Chr.7: 93215559-93215559 on Build GRCh38
4959985 c.633G>A p.K211K Chr.7: 93215483-93215483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757633841 Chr.7: 93215530-93215530 on Build GRCh38
rs752962182 Chr.7: 93215411-93215411 on Build GRCh38
rs754856674 Chr.7: 93215517-93215518 on Build GRCh38
rs140071737 Chr.7: 93215500-93215500 on Build GRCh38
rs774928451 Chr.7: 93215446-93215446 on Build GRCh38
rs758885062 Chr.7: 93215564-93215564 on Build GRCh38
rs996172648 Chr.7: 93215374-93215374 on Build GRCh38
rs767624363 Chr.7: 93215513-93215513 on Build GRCh38
rs754993085 Chr.7: 93215395-93215395 on Build GRCh38
rs953552013 Chr.7: 93215453-93215453 on Build GRCh38
rs779423772 Chr.7: 93215543-93215543 on Build GRCh38
rs544111598 Chr.7: 93215442-93215442 on Build GRCh38
rs893346874 Chr.7: 93215514-93215514 on Build GRCh38
rs779909896 Chr.7: 93215477-93215477 on Build GRCh38
rs749706675 Chr.7: 93215435-93215435 on Build GRCh38
rs750382824 Chr.7: 93215387-93215387 on Build GRCh38
rs757723703 Chr.7: 93215437-93215437 on Build GRCh38
rs754265690 Chr.7: 93215546-93215547 on Build GRCh38
rs761710225 Chr.7: 93215369-93215369 on Build GRCh38
rs750222743 Chr.7: 93215433-93215434 on Build GRCh38
rs753783130 Chr.7: 93215501-93215501 on Build GRCh38
rs776379052 Chr.7: 93215465-93215465 on Build GRCh38
rs764179545 Chr.7: 93215524-93215524 on Build GRCh38
rs761548759 Chr.7: 93215482-93215482 on Build GRCh38
rs371341619 Chr.7: 93215399-93215399 on Build GRCh38
rs764334445 Chr.7: 93215382-93215383 on Build GRCh38
rs765121250 Chr.7: 93215386-93215386 on Build GRCh38
rs199583781 Chr.7: 93215499-93215499 on Build GRCh38
rs763290229 Chr.7: 93215383-93215383 on Build GRCh38
rs1010167827 Chr.7: 93215502-93215502 on Build GRCh38
rs778175231 Chr.7: 93215416-93215416 on Build GRCh38
rs768234554 Chr.7: 93215458-93215458 on Build GRCh38
rs752764996 Chr.7: 93215363-93215364 on Build GRCh38
rs767503993 Chr.7: 93215407-93215407 on Build GRCh38
rs902095404 Chr.7: 93215364-93215364 on Build GRCh38
rs867151156 Chr.7: 93215516-93215517 on Build GRCh38
rs71107888 Chr.7: 93215391-93215391 on Build GRCh38
rs185525291 Chr.7: 93215398-93215398 on Build GRCh38
rs755911264 Chr.7: 93215445-93215445 on Build GRCh38
rs754130412 Chr.7: 93215526-93215526 on Build GRCh38
rs750961068 Chr.7: 93215552-93215552 on Build GRCh38
rs35608547 Chr.7: 93215506-93215506 on Build GRCh38
rs746390955 Chr.7: 93215438-93215438 on Build GRCh38
rs760917082 Chr.7: 93215520-93215520 on Build GRCh38
rs776345158 Chr.7: 93215362-93215362 on Build GRCh38

View Full Product Details