Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 48509970-48510243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRYL
Entrez Gene ID
285527
Gene Name
FRY like transcription coactivator
Gene Aliases
AF4p12, KIAA0826, MOR2
UniGene
Hs.595553
Gene Chromosome Location
Chr.4: 48497363-48780312 on Build GRCh38
Transcripts
NM_015030.1 XM_005248090.4 XM_005248093.4 XM_011513680.2 XM_011513681.2 XM_011513682.2 XM_011513683.2 XM_011513684.2 XM_017008046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
734276 c.8325C>G p.L2775L Chr.4: 48510128-48510128 on Build GRCh38
6667351 c.8323C>T p.L2775F Chr.4: 48510130-48510130 on Build GRCh38
1192274 c.8386G>A p.A2796T Chr.4: 48510067-48510067 on Build GRCh38
587358 c.8359G>A p.D2787N Chr.4: 48510094-48510094 on Build GRCh38
5031411 c.8302T>C p.S2768P Chr.4: 48510151-48510151 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370537290 Chr.4: 48510175-48510175 on Build GRCh38
rs747500062 Chr.4: 48510036-48510036 on Build GRCh38
rs771363059 Chr.4: 48510047-48510047 on Build GRCh38
rs756784787 Chr.4: 48510192-48510192 on Build GRCh38
rs775985306 Chr.4: 48510203-48510203 on Build GRCh38
rs181986849 Chr.4: 48510028-48510028 on Build GRCh38
rs75272765 Chr.4: 48510084-48510084 on Build GRCh38
rs770036701 Chr.4: 48510198-48510198 on Build GRCh38
rs761985834 Chr.4: 48510104-48510104 on Build GRCh38
rs753618579 Chr.4: 48510014-48510014 on Build GRCh38
rs775548133 Chr.4: 48510080-48510080 on Build GRCh38
rs753964830 Chr.4: 48510055-48510055 on Build GRCh38
rs757815929 Chr.4: 48510032-48510032 on Build GRCh38
rs754829406 Chr.4: 48510018-48510018 on Build GRCh38
rs564135770 Chr.4: 48510184-48510184 on Build GRCh38
rs73814779 Chr.4: 48510160-48510160 on Build GRCh38
rs777260003 Chr.4: 48510186-48510186 on Build GRCh38
rs532761015 Chr.4: 48510196-48510196 on Build GRCh38
rs747864906 Chr.4: 48510031-48510031 on Build GRCh38
rs770225988 Chr.4: 48510073-48510073 on Build GRCh38
rs199599571 Chr.4: 48510067-48510067 on Build GRCh38
rs113883732 Chr.4: 48510172-48510172 on Build GRCh38
rs777389969 Chr.4: 48510035-48510035 on Build GRCh38
rs1020807976 Chr.4: 48509993-48509993 on Build GRCh38
rs545935972 Chr.4: 48510169-48510169 on Build GRCh38
rs774642778 Chr.4: 48510103-48510103 on Build GRCh38
rs201844693 Chr.4: 48510152-48510152 on Build GRCh38
rs764070607 Chr.4: 48510089-48510089 on Build GRCh38
rs376997886 Chr.4: 48510071-48510071 on Build GRCh38
rs749469966 Chr.4: 48510207-48510207 on Build GRCh38
rs574639408 Chr.4: 48510005-48510005 on Build GRCh38
rs374603426 Chr.4: 48510191-48510191 on Build GRCh38

View Full Product Details