Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123605944-123606197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX55
Entrez Gene ID
57696
Gene Name
DEAD-box helicase 55
Gene Aliases
-
UniGene
Hs.286173
Gene Chromosome Location
Chr.12: 123602077-123620943 on Build GRCh38
Transcripts
NM_020936.2 NR_135104.1 NR_135105.1 XM_011538608.2 XM_017019710.1 XM_017019711.1 XM_017019712.1 XM_017019713.1 XM_017019714.1 XM_017019715.1 XM_017019716.1 XM_017019717.1 XM_017019718.1 XM_017019719.1 XM_017019720.1 XM_017019721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1359756 c.184G>A p.A62T Chr.12: 123606097-123606097 on Build GRCh38
3871001 c.200T>A p.I67N Chr.12: 123606113-123606113 on Build GRCh38
224442 c.159+1G>A p.? Chr.12: 123605982-123605982 on Build GRCh38
1746828 c.205G>T p.E69* Chr.12: 123606118-123606118 on Build GRCh38
7314660 c.160-8G>T p.? Chr.12: 123606065-123606065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577779871 Chr.12: 123606021-123606021 on Build GRCh38
rs768962905 Chr.12: 123606022-123606022 on Build GRCh38
rs779248003 Chr.12: 123606128-123606128 on Build GRCh38
rs398056034 Chr.12: 123606063-123606064 on Build GRCh38
rs970151108 Chr.12: 123606158-123606158 on Build GRCh38
rs762860675 Chr.12: 123606060-123606060 on Build GRCh38
rs756742204 Chr.12: 123606028-123606029 on Build GRCh38
rs148409263 Chr.12: 123606150-123606150 on Build GRCh38
rs775265531 Chr.12: 123606062-123606062 on Build GRCh38
rs780438032 Chr.12: 123606014-123606014 on Build GRCh38
rs777265413 Chr.12: 123606147-123606148 on Build GRCh38
rs559610605 Chr.12: 123606013-123606013 on Build GRCh38
rs748750646 Chr.12: 123606136-123606136 on Build GRCh38
rs777882464 Chr.12: 123605970-123605970 on Build GRCh38
rs201659723 Chr.12: 123606059-123606059 on Build GRCh38
rs769213957 Chr.12: 123606128-123606129 on Build GRCh38
rs889819160 Chr.12: 123605981-123605981 on Build GRCh38
rs892882894 Chr.12: 123606130-123606130 on Build GRCh38
rs776240783 Chr.12: 123606138-123606138 on Build GRCh38
rs767387538 Chr.12: 123606162-123606162 on Build GRCh38
rs761850100 Chr.12: 123606160-123606160 on Build GRCh38
rs368408656 Chr.12: 123606149-123606149 on Build GRCh38
rs540845812 Chr.12: 123605987-123605987 on Build GRCh38
rs780278642 Chr.12: 123606111-123606111 on Build GRCh38
rs4047521 Chr.12: 123606059-123606060 on Build GRCh38
rs745398122 Chr.12: 123606139-123606139 on Build GRCh38
rs758447593 Chr.12: 123605969-123605969 on Build GRCh38
rs774855791 Chr.12: 123606030-123606030 on Build GRCh38
rs769885138 Chr.12: 123605994-123605994 on Build GRCh38
rs770544787 Chr.12: 123606047-123606047 on Build GRCh38
rs746168787 Chr.12: 123605992-123605992 on Build GRCh38
rs762423517 Chr.12: 123606146-123606147 on Build GRCh38
rs1011764298 Chr.12: 123606145-123606145 on Build GRCh38
rs749287317 Chr.12: 123606029-123606035 on Build GRCh38
rs755342549 Chr.12: 123606124-123606124 on Build GRCh38
rs138847573 Chr.12: 123606096-123606096 on Build GRCh38
rs1045665944 Chr.12: 123606086-123606086 on Build GRCh38
rs138269355 Chr.12: 123606119-123606119 on Build GRCh38
rs767693727 Chr.12: 123606100-123606100 on Build GRCh38
rs116143975 Chr.12: 123606097-123606097 on Build GRCh38
rs752354140 Chr.12: 123606056-123606056 on Build GRCh38
rs7973689 Chr.12: 123606108-123606108 on Build GRCh38
rs750439210 Chr.12: 123606101-123606101 on Build GRCh38
rs768964196 Chr.12: 123606129-123606133 on Build GRCh38
rs762163278 Chr.12: 123606041-123606041 on Build GRCh38
rs113048238 Chr.12: 123606018-123606018 on Build GRCh38
rs751491547 Chr.12: 123605976-123605976 on Build GRCh38
rs201126940 Chr.12: 123605980-123605980 on Build GRCh38
rs545134794 Chr.12: 123606054-123606054 on Build GRCh38
rs775293345 Chr.12: 123606147-123606147 on Build GRCh38
rs772440374 Chr.12: 123606137-123606137 on Build GRCh38
rs375434150 Chr.12: 123606053-123606053 on Build GRCh38
rs778405409 Chr.12: 123606032-123606033 on Build GRCh38
rs771648048 Chr.12: 123606059-123606062 on Build GRCh38
rs774003506 Chr.12: 123606153-123606153 on Build GRCh38
rs199551284 Chr.12: 123606015-123606015 on Build GRCh38
rs763921238 Chr.12: 123606087-123606087 on Build GRCh38
rs116717261 Chr.12: 123606145-123606145 on Build GRCh38

View Full Product Details