Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50960929-50961154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIGD1C
Entrez Gene ID
613227
Gene Name
HIG1 hypoxia inducible domain family member 1C
Gene Aliases
GM921
UniGene
Hs.702849
Gene Chromosome Location
Chr.12: 50936223-50972592 on Build GRCh38
Transcripts
NM_001109619.1 XM_011538649.2 XM_017019783.1 XM_017019784.1 XM_017019785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1209671 c.154G>T p.D52Y Chr.12: 50961027-50961027 on Build GRCh38
4534012 c.203G>A p.G68E Chr.12: 50961076-50961076 on Build GRCh38
3998830 c.147C>T p.Y49Y Chr.12: 50961020-50961020 on Build GRCh38
3688257 c.186G>A p.M62I Chr.12: 50961059-50961059 on Build GRCh38
3359785 c.130C>G p.L44V Chr.12: 50961003-50961003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375581487 Chr.12: 50961114-50961114 on Build GRCh38
rs1012990764 Chr.12: 50961093-50961093 on Build GRCh38
rs755120419 Chr.12: 50960993-50960993 on Build GRCh38
rs371018971 Chr.12: 50961102-50961102 on Build GRCh38
rs145639855 Chr.12: 50961059-50961059 on Build GRCh38
rs758739336 Chr.12: 50961011-50961011 on Build GRCh38
rs765252671 Chr.12: 50961103-50961103 on Build GRCh38
rs1050589802 Chr.12: 50960971-50960971 on Build GRCh38
rs372681828 Chr.12: 50961037-50961037 on Build GRCh38
rs752870271 Chr.12: 50961109-50961109 on Build GRCh38
rs758799316 Chr.12: 50961110-50961110 on Build GRCh38
rs774279171 Chr.12: 50961073-50961073 on Build GRCh38
rs372097676 Chr.12: 50960961-50960961 on Build GRCh38
rs12306808 Chr.12: 50961020-50961020 on Build GRCh38
rs761754672 Chr.12: 50961092-50961092 on Build GRCh38
rs778412337 Chr.12: 50961112-50961112 on Build GRCh38
rs554369152 Chr.12: 50961059-50961060 on Build GRCh38
rs779407010 Chr.12: 50960994-50960994 on Build GRCh38
rs528933093 Chr.12: 50961055-50961055 on Build GRCh38
rs151336355 Chr.12: 50960959-50960959 on Build GRCh38
rs201612571 Chr.12: 50961099-50961099 on Build GRCh38
rs376053207 Chr.12: 50961116-50961116 on Build GRCh38
rs775305025 Chr.12: 50960956-50960956 on Build GRCh38
rs745554565 Chr.12: 50961120-50961120 on Build GRCh38
rs373079708 Chr.12: 50961064-50961064 on Build GRCh38
rs776325126 Chr.12: 50961056-50961056 on Build GRCh38
rs750613722 Chr.12: 50960966-50960966 on Build GRCh38
rs761603780 Chr.12: 50961057-50961057 on Build GRCh38
rs769300983 Chr.12: 50961122-50961122 on Build GRCh38
rs369825718 Chr.12: 50960977-50960977 on Build GRCh38
rs375956684 Chr.12: 50961045-50961045 on Build GRCh38
rs764144520 Chr.12: 50961070-50961070 on Build GRCh38
rs182661 Chr.12: 50961119-50961119 on Build GRCh38
rs199916635 Chr.12: 50961008-50961008 on Build GRCh38
rs140423925 Chr.12: 50960991-50960991 on Build GRCh38
rs751907670 Chr.12: 50961113-50961113 on Build GRCh38
rs374819592 Chr.12: 50960997-50960997 on Build GRCh38
rs765547056 Chr.12: 50961116-50961116 on Build GRCh38

View Full Product Details