Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154352164-154352435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
UniGene
Hs.195464
Gene Chromosome Location
Chr.X: 154348529-154374638 on Build GRCh38
Transcripts
NM_001110556.1 NM_001456.3 XM_011531127.1 XM_011531128.1 XM_011531129.1 XM_011531130.1 XM_011531131.1

SNP Detail

SNP ID Chromosome Location
rs876661183 Chr.X: 154352403-154352403 on Build GRCh38
rs1064822 Chr.X: 154352308-154352308 on Build GRCh38
rs797044738 Chr.X: 154352259-154352259 on Build GRCh38
rs782358643 Chr.X: 154352404-154352404 on Build GRCh38
rs782621818 Chr.X: 154352349-154352349 on Build GRCh38
rs782238357 Chr.X: 154352353-154352353 on Build GRCh38
rs11541268 Chr.X: 154352196-154352196 on Build GRCh38
rs782132724 Chr.X: 154352263-154352263 on Build GRCh38
rs782393623 Chr.X: 154352359-154352359 on Build GRCh38
rs782562592 Chr.X: 154352343-154352343 on Build GRCh38
rs782408769 Chr.X: 154352275-154352275 on Build GRCh38
rs782730629 Chr.X: 154352302-154352302 on Build GRCh38
rs782088697 Chr.X: 154352397-154352397 on Build GRCh38
rs782053982 Chr.X: 154352299-154352299 on Build GRCh38
rs782037508 Chr.X: 154352294-154352294 on Build GRCh38
rs113510895 Chr.X: 154352208-154352208 on Build GRCh38
rs1045173164 Chr.X: 154352376-154352376 on Build GRCh38
rs781801702 Chr.X: 154352344-154352344 on Build GRCh38
rs781935515 Chr.X: 154352337-154352337 on Build GRCh38
rs953454766 Chr.X: 154352222-154352222 on Build GRCh38
rs786205178 Chr.X: 154352312-154352315 on Build GRCh38
rs797045581 Chr.X: 154352231-154352231 on Build GRCh38
rs375499070 Chr.X: 154352338-154352338 on Build GRCh38
rs781984274 Chr.X: 154352225-154352225 on Build GRCh38
rs782299541 Chr.X: 154352379-154352379 on Build GRCh38
rs782053327 Chr.X: 154352386-154352386 on Build GRCh38
rs368547421 Chr.X: 154352348-154352348 on Build GRCh38
rs373913879 Chr.X: 154352200-154352200 on Build GRCh38
rs782314560 Chr.X: 154352380-154352380 on Build GRCh38
rs782352190 Chr.X: 154352212-154352212 on Build GRCh38
rs786205202 Chr.X: 154352370-154352370 on Build GRCh38
rs863223646 Chr.X: 154352349-154352349 on Build GRCh38
rs782658805 Chr.X: 154352365-154352365 on Build GRCh38
rs781992483 Chr.X: 154352410-154352410 on Build GRCh38

View Full Product Details