Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137052146-137052417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7448634 c.185A>T p.K62M Chr.9: 137052281-137052281 on Build GRCh38
4407395 c.118-4G>C p.? Chr.9: 137052352-137052352 on Build GRCh38
753064 c.221C>G p.S74C Chr.9: 137052245-137052245 on Build GRCh38
5938231 c.200G>A p.G67D Chr.9: 137052266-137052266 on Build GRCh38
1723073 c.118-4delG p.? Chr.9: 137052352-137052352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374786968 Chr.9: 137052373-137052373 on Build GRCh38
rs758457418 Chr.9: 137052386-137052386 on Build GRCh38
rs185233892 Chr.9: 137052203-137052203 on Build GRCh38
rs2271867 Chr.9: 137052352-137052352 on Build GRCh38
rs201777122 Chr.9: 137052331-137052331 on Build GRCh38
rs764690641 Chr.9: 137052253-137052253 on Build GRCh38
rs773897122 Chr.9: 137052380-137052380 on Build GRCh38
rs749352079 Chr.9: 137052201-137052201 on Build GRCh38
rs369220715 Chr.9: 137052385-137052385 on Build GRCh38
rs147614606 Chr.9: 137052317-137052317 on Build GRCh38
rs545396334 Chr.9: 137052351-137052351 on Build GRCh38
rs377158813 Chr.9: 137052290-137052290 on Build GRCh38
rs367730962 Chr.9: 137052202-137052202 on Build GRCh38
rs2271866 Chr.9: 137052313-137052313 on Build GRCh38
rs750237852 Chr.9: 137052389-137052389 on Build GRCh38
rs781084499 Chr.9: 137052329-137052329 on Build GRCh38
rs775767357 Chr.9: 137052197-137052197 on Build GRCh38
rs1006409689 Chr.9: 137052311-137052311 on Build GRCh38
rs372330901 Chr.9: 137052218-137052218 on Build GRCh38
rs747218051 Chr.9: 137052348-137052348 on Build GRCh38
rs756179116 Chr.9: 137052390-137052390 on Build GRCh38
rs144578226 Chr.9: 137052273-137052273 on Build GRCh38
rs766858896 Chr.9: 137052238-137052238 on Build GRCh38
rs140457830 Chr.9: 137052279-137052279 on Build GRCh38
rs575707189 Chr.9: 137052361-137052361 on Build GRCh38
rs773685339 Chr.9: 137052294-137052294 on Build GRCh38
rs145319457 Chr.9: 137052314-137052314 on Build GRCh38
rs767421632 Chr.9: 137052388-137052388 on Build GRCh38
rs748164984 Chr.9: 137052339-137052339 on Build GRCh38
rs752372010 Chr.9: 137052266-137052266 on Build GRCh38
rs200039691 Chr.9: 137052349-137052349 on Build GRCh38
rs763628632 Chr.9: 137052316-137052316 on Build GRCh38
rs753396274 Chr.9: 137052242-137052242 on Build GRCh38
rs770766766 Chr.9: 137052188-137052188 on Build GRCh38
rs772181440 Chr.9: 137052292-137052292 on Build GRCh38
rs766206680 Chr.9: 137052395-137052395 on Build GRCh38
rs775112000 Chr.9: 137052359-137052359 on Build GRCh38
rs762721975 Chr.9: 137052315-137052315 on Build GRCh38
rs756047018 Chr.9: 137052332-137052332 on Build GRCh38
rs759766401 Chr.9: 137052207-137052208 on Build GRCh38
rs777157838 Chr.9: 137052309-137052309 on Build GRCh38
rs752862041 Chr.9: 137052398-137052398 on Build GRCh38
rs1026575682 Chr.9: 137052271-137052271 on Build GRCh38
rs112297149 Chr.9: 137052215-137052215 on Build GRCh38
rs779203826 Chr.9: 137052338-137052338 on Build GRCh38
rs777688009 Chr.9: 137052270-137052270 on Build GRCh38
rs757932715 Chr.9: 137052267-137052267 on Build GRCh38
rs751294963 Chr.9: 137052272-137052272 on Build GRCh38
rs761312454 Chr.9: 137052232-137052232 on Build GRCh38
rs200817157 Chr.9: 137052386-137052386 on Build GRCh38
rs200217940 Chr.9: 137052289-137052289 on Build GRCh38
rs139779693 Chr.9: 137052274-137052274 on Build GRCh38
rs544337236 Chr.9: 137052365-137052365 on Build GRCh38
rs759020463 Chr.9: 137052250-137052250 on Build GRCh38
rs768878814 Chr.9: 137052280-137052280 on Build GRCh38
rs554571357 Chr.9: 137052359-137052359 on Build GRCh38
rs765375045 Chr.9: 137052350-137052351 on Build GRCh38
rs760925984 Chr.9: 137052295-137052295 on Build GRCh38
rs764707372 Chr.9: 137052391-137052398 on Build GRCh38
rs559838122 Chr.9: 137052330-137052330 on Build GRCh38
rs771471717 Chr.9: 137052302-137052302 on Build GRCh38
rs140533815 Chr.9: 137052340-137052340 on Build GRCh38
rs767935542 Chr.9: 137052217-137052217 on Build GRCh38
rs774744917 Chr.9: 137052207-137052207 on Build GRCh38
rs770269597 Chr.9: 137052353-137052353 on Build GRCh38

View Full Product Details