Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 47074115-47074388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYA2
Entrez Gene ID
2139
Gene Name
EYA transcriptional coactivator and phosphatase 2
Gene Aliases
EAB1
UniGene
Hs.472877
Gene Chromosome Location
Chr.20: 46894624-47188844 on Build GRCh38
Transcripts
NM_005244.4 NM_172110.3 XM_005260327.2 XM_017027721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232276 c.584C>T p.S195L Chr.20: 47074258-47074258 on Build GRCh38
3547333 c.648C>T p.S216S Chr.20: 47074322-47074322 on Build GRCh38
3547332 c.524C>T p.P175L Chr.20: 47074198-47074198 on Build GRCh38
724365 c.509C>A p.P170H Chr.20: 47074183-47074183 on Build GRCh38
478238 c.580C>A p.P194T Chr.20: 47074254-47074254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770631317 Chr.20: 47074221-47074221 on Build GRCh38
rs149701901 Chr.20: 47074208-47074208 on Build GRCh38
rs1033490626 Chr.20: 47074190-47074190 on Build GRCh38
rs991612630 Chr.20: 47074198-47074198 on Build GRCh38
rs578044921 Chr.20: 47074281-47074281 on Build GRCh38
rs760087651 Chr.20: 47074302-47074302 on Build GRCh38
rs761438426 Chr.20: 47074274-47074274 on Build GRCh38
rs1015220349 Chr.20: 47074323-47074323 on Build GRCh38
rs202115929 Chr.20: 47074233-47074233 on Build GRCh38
rs752897315 Chr.20: 47074231-47074231 on Build GRCh38
rs764206024 Chr.20: 47074307-47074307 on Build GRCh38
rs768036699 Chr.20: 47074303-47074303 on Build GRCh38
rs374467565 Chr.20: 47074271-47074271 on Build GRCh38
rs781279227 Chr.20: 47074351-47074351 on Build GRCh38
rs933557936 Chr.20: 47074240-47074240 on Build GRCh38
rs772932365 Chr.20: 47074279-47074279 on Build GRCh38
rs775507043 Chr.20: 47074167-47074167 on Build GRCh38
rs759901684 Chr.20: 47074342-47074344 on Build GRCh38
rs553735931 Chr.20: 47074266-47074266 on Build GRCh38
rs769487812 Chr.20: 47074365-47074365 on Build GRCh38
rs776388587 Chr.20: 47074176-47074176 on Build GRCh38
rs376496454 Chr.20: 47074238-47074238 on Build GRCh38
rs771868565 Chr.20: 47074155-47074155 on Build GRCh38
rs754151844 Chr.20: 47074308-47074308 on Build GRCh38
rs772736202 Chr.20: 47074367-47074367 on Build GRCh38
rs779848203 Chr.20: 47074255-47074255 on Build GRCh38
rs866678281 Chr.20: 47074321-47074321 on Build GRCh38
rs761605751 Chr.20: 47074194-47074194 on Build GRCh38
rs747603278 Chr.20: 47074361-47074361 on Build GRCh38
rs149474635 Chr.20: 47074185-47074185 on Build GRCh38
rs945421711 Chr.20: 47074206-47074206 on Build GRCh38
rs766738869 Chr.20: 47074209-47074209 on Build GRCh38
rs371083777 Chr.20: 47074258-47074258 on Build GRCh38
rs535313513 Chr.20: 47074264-47074264 on Build GRCh38
rs142696386 Chr.20: 47074305-47074305 on Build GRCh38
rs746820594 Chr.20: 47074348-47074348 on Build GRCh38
rs750713751 Chr.20: 47074199-47074199 on Build GRCh38
rs140248088 Chr.20: 47074237-47074237 on Build GRCh38
rs370935322 Chr.20: 47074211-47074211 on Build GRCh38
rs765002260 Chr.20: 47074197-47074197 on Build GRCh38
rs1013194543 Chr.20: 47074262-47074262 on Build GRCh38
rs781290141 Chr.20: 47074225-47074225 on Build GRCh38
rs945048393 Chr.20: 47074342-47074342 on Build GRCh38
rs754924338 Chr.20: 47074350-47074350 on Build GRCh38
rs910742652 Chr.20: 47074269-47074269 on Build GRCh38
rs766835345 Chr.20: 47074296-47074296 on Build GRCh38
rs769504030 Chr.20: 47074277-47074277 on Build GRCh38
rs377702972 Chr.20: 47074175-47074175 on Build GRCh38
rs752737822 Chr.20: 47074304-47074304 on Build GRCh38
rs756282686 Chr.20: 47074232-47074232 on Build GRCh38
rs774691281 Chr.20: 47074299-47074299 on Build GRCh38
rs372882937 Chr.20: 47074150-47074150 on Build GRCh38
rs779945803 Chr.20: 47074344-47074344 on Build GRCh38
rs760086878 Chr.20: 47074172-47074172 on Build GRCh38
rs367734889 Chr.20: 47074322-47074322 on Build GRCh38
rs569736987 Chr.20: 47074196-47074196 on Build GRCh38
rs776212689 Chr.20: 47074228-47074275 on Build GRCh38

View Full Product Details