Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69371434-69371695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189A2
Entrez Gene ID
9413
Gene Name
family with sequence similarity 189 member A2
Gene Aliases
C9orf61, X123
UniGene
Hs.118003
Gene Chromosome Location
Chr.9: 69324572-69392461 on Build GRCh38
Transcripts
NM_001127608.1 NM_004816.3 XM_005252307.4 XM_017015324.1 XM_017015325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4849965 c.58C>G p.L20V Chr.9: 69371544-69371544 on Build GRCh38
1462760 c.73C>A p.L25I Chr.9: 69371559-69371559 on Build GRCh38
1643858 c.115T>C p.C39R Chr.9: 69371601-69371601 on Build GRCh38
5120121 c.13-3C>A p.? Chr.9: 69371496-69371496 on Build GRCh38
3730106 c.123+3A>T p.? Chr.9: 69371612-69371612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758864301 Chr.9: 69371589-69371589 on Build GRCh38
rs539569315 Chr.9: 69371524-69371524 on Build GRCh38
rs184081860 Chr.9: 69371592-69371592 on Build GRCh38
rs377735148 Chr.9: 69371634-69371634 on Build GRCh38
rs533908108 Chr.9: 69371632-69371632 on Build GRCh38
rs555831386 Chr.9: 69371633-69371633 on Build GRCh38
rs760904923 Chr.9: 69371519-69371519 on Build GRCh38
rs926362339 Chr.9: 69371673-69371673 on Build GRCh38
rs112699608 Chr.9: 69371653-69371653 on Build GRCh38
rs753952590 Chr.9: 69371532-69371532 on Build GRCh38
rs770748554 Chr.9: 69371607-69371607 on Build GRCh38
rs571310154 Chr.9: 69371515-69371515 on Build GRCh38
rs140797265 Chr.9: 69371555-69371555 on Build GRCh38
rs753258017 Chr.9: 69371609-69371609 on Build GRCh38
rs984587878 Chr.9: 69371485-69371485 on Build GRCh38
rs144772522 Chr.9: 69371606-69371606 on Build GRCh38
rs1037660190 Chr.9: 69371518-69371518 on Build GRCh38
rs867626649 Chr.9: 69371568-69371569 on Build GRCh38
rs1016171114 Chr.9: 69371507-69371507 on Build GRCh38
rs757677212 Chr.9: 69371538-69371538 on Build GRCh38
rs779524519 Chr.9: 69371548-69371548 on Build GRCh38
rs777376133 Chr.9: 69371604-69371604 on Build GRCh38
rs780287782 Chr.9: 69371591-69371591 on Build GRCh38
rs769592488 Chr.9: 69371597-69371597 on Build GRCh38
rs201643409 Chr.9: 69371480-69371480 on Build GRCh38
rs970889514 Chr.9: 69371616-69371616 on Build GRCh38
rs552779903 Chr.9: 69371655-69371655 on Build GRCh38
rs374296854 Chr.9: 69371501-69371501 on Build GRCh38
rs759422185 Chr.9: 69371492-69371492 on Build GRCh38
rs750876540 Chr.9: 69371660-69371660 on Build GRCh38
rs372581278 Chr.9: 69371618-69371618 on Build GRCh38
rs12001021 Chr.9: 69371672-69371672 on Build GRCh38
rs764114694 Chr.9: 69371530-69371530 on Build GRCh38
rs773983206 Chr.9: 69371491-69371491 on Build GRCh38
rs764341810 Chr.9: 69371635-69371635 on Build GRCh38
rs753896525 Chr.9: 69371640-69371640 on Build GRCh38
rs865986417 Chr.9: 69371567-69371568 on Build GRCh38

View Full Product Details