Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70366572-70366817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNMB4
Entrez Gene ID
27345
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 4
Gene Aliases
-
UniGene
Hs.525529
Gene Chromosome Location
Chr.12: 70366220-70434292 on Build GRCh38
Transcripts
NM_014505.5 XM_011538188.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377566 c.55C>T p.R19W Chr.12: 70366789-70366789 on Build GRCh38
3464716 c.25G>A p.E9K Chr.12: 70366759-70366759 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748946457 Chr.12: 70366723-70366723 on Build GRCh38
rs780649197 Chr.12: 70366716-70366716 on Build GRCh38
rs905208972 Chr.12: 70366600-70366600 on Build GRCh38
rs775402368 Chr.12: 70366733-70366733 on Build GRCh38
rs1030236200 Chr.12: 70366705-70366705 on Build GRCh38
rs753525779 Chr.12: 70366747-70366747 on Build GRCh38
rs551085353 Chr.12: 70366721-70366721 on Build GRCh38
rs765309715 Chr.12: 70366777-70366777 on Build GRCh38
rs777327993 Chr.12: 70366722-70366722 on Build GRCh38
rs757331385 Chr.12: 70366685-70366685 on Build GRCh38
rs758535497 Chr.12: 70366791-70366791 on Build GRCh38
rs772056073 Chr.12: 70366730-70366730 on Build GRCh38
rs1055177787 Chr.12: 70366699-70366699 on Build GRCh38
rs752078274 Chr.12: 70366794-70366794 on Build GRCh38
rs35578708 Chr.12: 70366639-70366640 on Build GRCh38
rs1053886258 Chr.12: 70366639-70366639 on Build GRCh38
rs750428962 Chr.12: 70366708-70366708 on Build GRCh38
rs762125328 Chr.12: 70366766-70366766 on Build GRCh38
rs1007495741 Chr.12: 70366725-70366726 on Build GRCh38
rs532514417 Chr.12: 70366706-70366706 on Build GRCh38
rs1009444132 Chr.12: 70366673-70366673 on Build GRCh38
rs760504525 Chr.12: 70366744-70366744 on Build GRCh38
rs763928883 Chr.12: 70366746-70366746 on Build GRCh38
rs1001320332 Chr.12: 70366625-70366626 on Build GRCh38
rs887809772 Chr.12: 70366689-70366689 on Build GRCh38
rs370331603 Chr.12: 70366758-70366758 on Build GRCh38
rs112372380 Chr.12: 70366727-70366727 on Build GRCh38
rs372457321 Chr.12: 70366717-70366717 on Build GRCh38
rs779070594 Chr.12: 70366693-70366693 on Build GRCh38
rs565254738 Chr.12: 70366646-70366646 on Build GRCh38
rs536892206 Chr.12: 70366784-70366784 on Build GRCh38

View Full Product Details