Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99699982-99700229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP46A1
Entrez Gene ID
10858
Gene Name
cytochrome P450 family 46 subfamily A member 1
Gene Aliases
CP46, CYP46
UniGene
Hs.25121
Gene Chromosome Location
Chr.14: 99684262-99727321 on Build GRCh38
Transcripts
NM_006668.1 XM_011536364.1 XM_017020933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249600 c.380C>G p.S127C Chr.14: 99700038-99700038 on Build GRCh38
1678247 c.430G>A p.A144T Chr.14: 99700088-99700088 on Build GRCh38
3814227 c.382G>A p.E128K Chr.14: 99700040-99700040 on Build GRCh38
6639322 c.397C>T p.R133C Chr.14: 99700055-99700055 on Build GRCh38
1493148 c.440G>A p.R147Q Chr.14: 99700098-99700098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892620906 Chr.14: 99700175-99700175 on Build GRCh38
rs563389722 Chr.14: 99700153-99700153 on Build GRCh38
rs747142169 Chr.14: 99700009-99700009 on Build GRCh38
rs540821243 Chr.14: 99700113-99700113 on Build GRCh38
rs1004562965 Chr.14: 99700203-99700203 on Build GRCh38
rs781018972 Chr.14: 99700127-99700127 on Build GRCh38
rs765829489 Chr.14: 99700101-99700101 on Build GRCh38
rs754400703 Chr.14: 99700108-99700108 on Build GRCh38
rs371296881 Chr.14: 99700135-99700135 on Build GRCh38
rs144966483 Chr.14: 99700036-99700036 on Build GRCh38
rs765667580 Chr.14: 99700114-99700114 on Build GRCh38
rs199681773 Chr.14: 99700011-99700011 on Build GRCh38
rs201808768 Chr.14: 99700129-99700129 on Build GRCh38
rs139245529 Chr.14: 99700039-99700039 on Build GRCh38
rs761460288 Chr.14: 99700054-99700054 on Build GRCh38
rs904875341 Chr.14: 99700199-99700199 on Build GRCh38
rs745873211 Chr.14: 99700136-99700136 on Build GRCh38
rs758765792 Chr.14: 99700126-99700126 on Build GRCh38
rs1039917206 Chr.14: 99700121-99700121 on Build GRCh38
rs199575465 Chr.14: 99700116-99700116 on Build GRCh38
rs780187016 Chr.14: 99700021-99700021 on Build GRCh38
rs376737456 Chr.14: 99700122-99700122 on Build GRCh38
rs1013844758 Chr.14: 99700186-99700186 on Build GRCh38
rs772703431 Chr.14: 99700082-99700082 on Build GRCh38
rs771783206 Chr.14: 99700081-99700081 on Build GRCh38
rs754975646 Chr.14: 99700140-99700140 on Build GRCh38
rs1051114882 Chr.14: 99700104-99700104 on Build GRCh38
rs751708118 Chr.14: 99700123-99700123 on Build GRCh38
rs760275943 Chr.14: 99700097-99700097 on Build GRCh38
rs931948427 Chr.14: 99700107-99700107 on Build GRCh38
rs1025607871 Chr.14: 99700195-99700195 on Build GRCh38
rs774289118 Chr.14: 99700040-99700040 on Build GRCh38
rs529240332 Chr.14: 99700158-99700158 on Build GRCh38
rs943880 Chr.14: 99700152-99700152 on Build GRCh38

View Full Product Details