Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176643686-176643921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E1B
Entrez Gene ID
253314
Gene Name
eukaryotic translation initiation factor 4E family member 1B
Gene Aliases
-
UniGene
Hs.448362
Gene Chromosome Location
Chr.5: 176630613-176646644 on Build GRCh38
Transcripts
NM_001099408.1 XM_011534499.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6470210 c.255G>A p.L85L Chr.5: 176643693-176643693 on Build GRCh38
3853878 c.296+1G>A p.? Chr.5: 176643735-176643735 on Build GRCh38
4934837 c.253C>G p.L85V Chr.5: 176643691-176643691 on Build GRCh38
3365992 c.271G>A p.V91M Chr.5: 176643709-176643709 on Build GRCh38
338353 c.280G>A p.V94M Chr.5: 176643718-176643718 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974078518 Chr.5: 176643835-176643835 on Build GRCh38
rs376452636 Chr.5: 176643872-176643872 on Build GRCh38
rs754129447 Chr.5: 176643766-176643768 on Build GRCh38
rs568253648 Chr.5: 176643883-176643883 on Build GRCh38
rs946437043 Chr.5: 176643798-176643798 on Build GRCh38
rs535606524 Chr.5: 176643887-176643887 on Build GRCh38
rs184522665 Chr.5: 176643797-176643797 on Build GRCh38
rs1025245757 Chr.5: 176643803-176643803 on Build GRCh38
rs766435727 Chr.5: 176643745-176643745 on Build GRCh38
rs765558809 Chr.5: 176643724-176643724 on Build GRCh38
rs565788684 Chr.5: 176643737-176643737 on Build GRCh38
rs574266946 Chr.5: 176643752-176643752 on Build GRCh38
rs760507724 Chr.5: 176643723-176643723 on Build GRCh38
rs756173276 Chr.5: 176643754-176643755 on Build GRCh38
rs1044835427 Chr.5: 176643809-176643809 on Build GRCh38
rs143987075 Chr.5: 176643830-176643830 on Build GRCh38
rs983259228 Chr.5: 176643807-176643807 on Build GRCh38
rs368052053 Chr.5: 176643773-176643773 on Build GRCh38
rs745323634 Chr.5: 176643783-176643783 on Build GRCh38
rs772168290 Chr.5: 176643784-176643784 on Build GRCh38
rs932916082 Chr.5: 176643848-176643848 on Build GRCh38
rs779979324 Chr.5: 176643765-176643766 on Build GRCh38
rs1051704836 Chr.5: 176643744-176643744 on Build GRCh38
rs898105627 Chr.5: 176643880-176643880 on Build GRCh38
rs778336353 Chr.5: 176643778-176643778 on Build GRCh38
rs905327595 Chr.5: 176643820-176643820 on Build GRCh38
rs750502319 Chr.5: 176643738-176643750 on Build GRCh38
rs1009746251 Chr.5: 176643757-176643757 on Build GRCh38
rs770591432 Chr.5: 176643777-176643777 on Build GRCh38
rs372658671 Chr.5: 176643791-176643791 on Build GRCh38
rs561940055 Chr.5: 176643708-176643708 on Build GRCh38
rs1000524445 Chr.5: 176643786-176643786 on Build GRCh38
rs932802977 Chr.5: 176643743-176643743 on Build GRCh38
rs200108394 Chr.5: 176643832-176643832 on Build GRCh38
rs779309244 Chr.5: 176643783-176643784 on Build GRCh38
rs533488547 Chr.5: 176643749-176643749 on Build GRCh38
rs755189174 Chr.5: 176643771-176643777 on Build GRCh38
rs547594222 Chr.5: 176643720-176643720 on Build GRCh38
rs756415431 Chr.5: 176643761-176643761 on Build GRCh38
rs1056721809 Chr.5: 176643869-176643869 on Build GRCh38
rs116092226 Chr.5: 176643714-176643714 on Build GRCh38
rs750551455 Chr.5: 176643734-176643734 on Build GRCh38
rs778189705 Chr.5: 176643770-176643770 on Build GRCh38
rs556468449 Chr.5: 176643867-176643867 on Build GRCh38
rs557333631 Chr.5: 176643735-176643735 on Build GRCh38
rs753089384 Chr.5: 176643756-176643756 on Build GRCh38
rs773912010 Chr.5: 176643712-176643712 on Build GRCh38
rs117853394 Chr.5: 176643841-176643841 on Build GRCh38
rs772113105 Chr.5: 176643719-176643719 on Build GRCh38

View Full Product Details