Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48962699-48962924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSHR
Entrez Gene ID
2492
Gene Name
follicle stimulating hormone receptor
Gene Aliases
FSHR1, FSHRO, LGR1, ODG1
UniGene
Hs.1428
Gene Chromosome Location
Chr.2: 48953161-49154527 on Build GRCh38
Transcripts
NM_000145.3 NM_181446.2 XM_011532733.2 XM_011532734.2 XM_011532735.2 XM_011532736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4894999 c.1914C>T p.F638F Chr.2: 48962907-48962907 on Build GRCh38
3426504 c.1906G>A p.D636N Chr.2: 48962915-48962915 on Build GRCh38
4094518 c.1922T>C p.L641P Chr.2: 48962899-48962899 on Build GRCh38
400568 c.1952C>T p.A651V Chr.2: 48962869-48962869 on Build GRCh38
5725383 c.1904G>A p.R635K Chr.2: 48962917-48962917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937425277 Chr.2: 48962859-48962859 on Build GRCh38
rs774725838 Chr.2: 48962835-48962835 on Build GRCh38
rs775980862 Chr.2: 48962753-48962753 on Build GRCh38
rs776161444 Chr.2: 48962857-48962857 on Build GRCh38
rs753348338 Chr.2: 48962767-48962767 on Build GRCh38
rs765968933 Chr.2: 48962760-48962760 on Build GRCh38
rs751333023 Chr.2: 48962888-48962888 on Build GRCh38
rs745960881 Chr.2: 48962735-48962735 on Build GRCh38
rs759315502 Chr.2: 48962862-48962862 on Build GRCh38
rs758214962 Chr.2: 48962849-48962849 on Build GRCh38
rs760351580 Chr.2: 48962757-48962757 on Build GRCh38
rs778929695 Chr.2: 48962794-48962794 on Build GRCh38
rs777591569 Chr.2: 48962788-48962788 on Build GRCh38
rs377657419 Chr.2: 48962850-48962850 on Build GRCh38
rs916438930 Chr.2: 48962829-48962829 on Build GRCh38
rs769877026 Chr.2: 48962808-48962808 on Build GRCh38
rs758390491 Chr.2: 48962877-48962877 on Build GRCh38
rs781418151 Chr.2: 48962896-48962896 on Build GRCh38
rs6166 Chr.2: 48962782-48962782 on Build GRCh38
rs749792082 Chr.2: 48962822-48962822 on Build GRCh38
rs112937052 Chr.2: 48962832-48962832 on Build GRCh38
rs978748810 Chr.2: 48962793-48962793 on Build GRCh38
rs775606372 Chr.2: 48962810-48962810 on Build GRCh38
rs754539288 Chr.2: 48962780-48962780 on Build GRCh38
rs12620825 Chr.2: 48962828-48962828 on Build GRCh38
rs756221634 Chr.2: 48962785-48962785 on Build GRCh38
rs752516112 Chr.2: 48962799-48962799 on Build GRCh38
rs199948641 Chr.2: 48962821-48962821 on Build GRCh38
rs764832498 Chr.2: 48962863-48962863 on Build GRCh38
rs547208879 Chr.2: 48962755-48962755 on Build GRCh38
rs375279102 Chr.2: 48962892-48962892 on Build GRCh38
rs752664767 Chr.2: 48962783-48962783 on Build GRCh38
rs781304663 Chr.2: 48962795-48962795 on Build GRCh38
rs148626637 Chr.2: 48962848-48962848 on Build GRCh38
rs201115329 Chr.2: 48962891-48962891 on Build GRCh38
rs745885725 Chr.2: 48962797-48962797 on Build GRCh38
rs147439856 Chr.2: 48962791-48962791 on Build GRCh38
rs752292800 Chr.2: 48962874-48962874 on Build GRCh38
rs918822982 Chr.2: 48962752-48962752 on Build GRCh38
rs984242436 Chr.2: 48962817-48962817 on Build GRCh38
rs774648218 Chr.2: 48962756-48962756 on Build GRCh38
rs763964308 Chr.2: 48962880-48962880 on Build GRCh38
rs149502612 Chr.2: 48962804-48962804 on Build GRCh38

View Full Product Details