Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128462470-128462697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETS1
Entrez Gene ID
2113
Gene Name
ETS proto-oncogene 1, transcription factor
Gene Aliases
ETS-1, EWSR2, c-ets-1, p54
UniGene
Hs.369438
Gene Chromosome Location
Chr.11: 128458761-128587593 on Build GRCh38
Transcripts
NM_001143820.1 NM_001162422.1 NM_005238.3 XM_011542649.2 XM_011542650.1 XM_011542651.1 XM_011542652.1 XM_017017314.1 XM_017017315.1 XM_017017316.1 XM_017017317.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3445160 c.1307G>A p.G436D Chr.11: 128462512-128462512 on Build GRCh38
5367813 c.938_939insA p.N313fs*18 Chr.11: 128462487-128462488 on Build GRCh38
4681822 c.1199delA p.N400fs*>42 Chr.11: 128462488-128462488 on Build GRCh38
5950161 c.927T>G p.Y309* Chr.11: 128462499-128462499 on Build GRCh38
5456092 c.930C>T p.Y310Y Chr.11: 128462496-128462496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773267009 Chr.11: 128462605-128462605 on Build GRCh38
rs201484607 Chr.11: 128462558-128462558 on Build GRCh38
rs917049327 Chr.11: 128462672-128462672 on Build GRCh38
rs1011979025 Chr.11: 128462656-128462656 on Build GRCh38
rs759600432 Chr.11: 128462608-128462608 on Build GRCh38
rs771714129 Chr.11: 128462594-128462597 on Build GRCh38
rs116502295 Chr.11: 128462620-128462620 on Build GRCh38
rs747880867 Chr.11: 128462592-128462592 on Build GRCh38
rs1034712187 Chr.11: 128462630-128462630 on Build GRCh38
rs199698372 Chr.11: 128462496-128462496 on Build GRCh38
rs781161246 Chr.11: 128462591-128462591 on Build GRCh38
rs779615012 Chr.11: 128462568-128462568 on Build GRCh38
rs772159155 Chr.11: 128462610-128462610 on Build GRCh38
rs775084273 Chr.11: 128462600-128462602 on Build GRCh38
rs746655453 Chr.11: 128462585-128462585 on Build GRCh38
rs757924801 Chr.11: 128462510-128462510 on Build GRCh38
rs369222024 Chr.11: 128462619-128462619 on Build GRCh38
rs769865019 Chr.11: 128462597-128462597 on Build GRCh38
rs992649300 Chr.11: 128462667-128462667 on Build GRCh38
rs768148563 Chr.11: 128462586-128462586 on Build GRCh38
rs959091286 Chr.11: 128462631-128462631 on Build GRCh38
rs762154390 Chr.11: 128462621-128462621 on Build GRCh38

View Full Product Details