Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41941105-41941362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO4
Entrez Gene ID
26272
Gene Name
F-box protein 4
Gene Aliases
FBX4
UniGene
Hs.165575
Gene Chromosome Location
Chr.5: 41925249-42040230 on Build GRCh38
Transcripts
NM_001297437.1 NM_012176.2 XM_006714470.3 XM_011514026.2 XM_011514027.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6343851 c.1125G>T p.E375D Chr.5: 41941242-41941242 on Build GRCh38
3342420 c.1118G>A p.G373D Chr.5: 41941235-41941235 on Build GRCh38
4141825 c.1075-8A>G p.? Chr.5: 41941184-41941184 on Build GRCh38
1068342 c.1132C>A p.L378I Chr.5: 41941249-41941249 on Build GRCh38
3616666 c.1163G>T p.*388L Chr.5: 41941280-41941280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747585526 Chr.5: 41941216-41941216 on Build GRCh38
rs1037924673 Chr.5: 41941338-41941338 on Build GRCh38
rs755292420 Chr.5: 41941189-41941189 on Build GRCh38
rs377698302 Chr.5: 41941154-41941154 on Build GRCh38
rs753947700 Chr.5: 41941188-41941188 on Build GRCh38
rs149145775 Chr.5: 41941248-41941248 on Build GRCh38
rs778334720 Chr.5: 41941148-41941148 on Build GRCh38
rs763048667 Chr.5: 41941330-41941330 on Build GRCh38
rs754477373 Chr.5: 41941286-41941286 on Build GRCh38
rs370478423 Chr.5: 41941231-41941231 on Build GRCh38
rs761587767 Chr.5: 41941166-41941166 on Build GRCh38
rs747740555 Chr.5: 41941271-41941271 on Build GRCh38
rs774824423 Chr.5: 41941161-41941161 on Build GRCh38
rs370657056 Chr.5: 41941321-41941321 on Build GRCh38
rs919463487 Chr.5: 41941315-41941315 on Build GRCh38
rs769354077 Chr.5: 41941159-41941159 on Build GRCh38
rs757658813 Chr.5: 41941223-41941223 on Build GRCh38
rs772842372 Chr.5: 41941291-41941291 on Build GRCh38
rs753171422 Chr.5: 41941194-41941194 on Build GRCh38
rs777937216 Chr.5: 41941208-41941208 on Build GRCh38
rs765552737 Chr.5: 41941191-41941191 on Build GRCh38
rs564998475 Chr.5: 41941259-41941259 on Build GRCh38
rs992855016 Chr.5: 41941280-41941280 on Build GRCh38
rs752977325 Chr.5: 41941328-41941328 on Build GRCh38
rs532213620 Chr.5: 41941316-41941316 on Build GRCh38
rs767412193 Chr.5: 41941167-41941167 on Build GRCh38
rs893525729 Chr.5: 41941135-41941135 on Build GRCh38
rs758851440 Chr.5: 41941207-41941207 on Build GRCh38
rs773214530 Chr.5: 41941179-41941179 on Build GRCh38
rs780153987 Chr.5: 41941164-41941164 on Build GRCh38
rs764544972 Chr.5: 41941332-41941332 on Build GRCh38
rs760717501 Chr.5: 41941311-41941311 on Build GRCh38
rs972541745 Chr.5: 41941264-41941264 on Build GRCh38
rs528126288 Chr.5: 41941242-41941242 on Build GRCh38
rs368508641 Chr.5: 41941140-41941140 on Build GRCh38
rs372987274 Chr.5: 41941181-41941181 on Build GRCh38
rs966559253 Chr.5: 41941150-41941150 on Build GRCh38
rs376695334 Chr.5: 41941270-41941270 on Build GRCh38
rs766510175 Chr.5: 41941187-41941187 on Build GRCh38
rs765353971 Chr.5: 41941327-41941327 on Build GRCh38
rs200501922 Chr.5: 41941162-41941162 on Build GRCh38

View Full Product Details