Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 49017374-49017631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSHR
Entrez Gene ID
2492
Gene Name
follicle stimulating hormone receptor
Gene Aliases
FSHR1, FSHRO, LGR1, ODG1
UniGene
Hs.1428
Gene Chromosome Location
Chr.2: 48953161-49154527 on Build GRCh38
Transcripts
NM_000145.3 NM_181446.2 XM_011532733.2 XM_011532734.2 XM_011532740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1327044 c.342G>C p.E114D Chr.2: 49017521-49017521 on Build GRCh38
3771105 c.307G>A p.E103K Chr.2: 49017556-49017556 on Build GRCh38
6542575 c.302G>A p.R101K Chr.2: 49017561-49017561 on Build GRCh38
6667487 c.339T>G p.P113P Chr.2: 49017524-49017524 on Build GRCh38
6667489 c.318C>G p.N106K Chr.2: 49017545-49017545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138344938 Chr.2: 49017538-49017538 on Build GRCh38
rs748145877 Chr.2: 49017456-49017456 on Build GRCh38
rs1006020716 Chr.2: 49017480-49017480 on Build GRCh38
rs760896533 Chr.2: 49017465-49017465 on Build GRCh38
rs375700282 Chr.2: 49017448-49017448 on Build GRCh38
rs141135052 Chr.2: 49017509-49017509 on Build GRCh38
rs368345067 Chr.2: 49017455-49017455 on Build GRCh38
rs375603115 Chr.2: 49017582-49017582 on Build GRCh38
rs753293607 Chr.2: 49017504-49017504 on Build GRCh38
rs547762464 Chr.2: 49017408-49017408 on Build GRCh38
rs768517310 Chr.2: 49017446-49017446 on Build GRCh38
rs771137333 Chr.2: 49017535-49017535 on Build GRCh38
rs997096162 Chr.2: 49017413-49017413 on Build GRCh38
rs201909194 Chr.2: 49017529-49017529 on Build GRCh38
rs565972285 Chr.2: 49017409-49017409 on Build GRCh38
rs199897274 Chr.2: 49017470-49017470 on Build GRCh38
rs746133198 Chr.2: 49017542-49017542 on Build GRCh38
rs947067242 Chr.2: 49017404-49017404 on Build GRCh38
rs751882320 Chr.2: 49017485-49017485 on Build GRCh38
rs771865054 Chr.2: 49017457-49017457 on Build GRCh38
rs767914211 Chr.2: 49017596-49017596 on Build GRCh38
rs1015650924 Chr.2: 49017461-49017461 on Build GRCh38
rs200915059 Chr.2: 49017524-49017524 on Build GRCh38
rs967494598 Chr.2: 49017402-49017402 on Build GRCh38
rs760697660 Chr.2: 49017526-49017526 on Build GRCh38
rs138079934 Chr.2: 49017496-49017496 on Build GRCh38
rs1126714 Chr.2: 49017528-49017528 on Build GRCh38
rs774057514 Chr.2: 49017530-49017530 on Build GRCh38
rs147355964 Chr.2: 49017551-49017551 on Build GRCh38
rs775769692 Chr.2: 49017559-49017559 on Build GRCh38
rs774735687 Chr.2: 49017585-49017585 on Build GRCh38
rs755772178 Chr.2: 49017443-49017443 on Build GRCh38
rs772925869 Chr.2: 49017464-49017464 on Build GRCh38
rs377596422 Chr.2: 49017445-49017445 on Build GRCh38
rs899820797 Chr.2: 49017482-49017482 on Build GRCh38
rs764510894 Chr.2: 49017584-49017584 on Build GRCh38
rs143972148 Chr.2: 49017501-49017501 on Build GRCh38
rs776814907 Chr.2: 49017468-49017468 on Build GRCh38
rs536713398 Chr.2: 49017508-49017508 on Build GRCh38
rs762103961 Chr.2: 49017586-49017586 on Build GRCh38
rs764505936 Chr.2: 49017481-49017481 on Build GRCh38
rs543592760 Chr.2: 49017572-49017572 on Build GRCh38
rs554447061 Chr.2: 49017469-49017469 on Build GRCh38
rs369119142 Chr.2: 49017599-49017599 on Build GRCh38
rs2091787 Chr.2: 49017420-49017420 on Build GRCh38
rs886056151 Chr.2: 49017490-49017490 on Build GRCh38
rs1028910072 Chr.2: 49017527-49017527 on Build GRCh38
rs763437371 Chr.2: 49017476-49017476 on Build GRCh38

View Full Product Details