Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82059859-82060132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS1L
Entrez Gene ID
64118
Gene Name
dihydrouridine synthase 1 like
Gene Aliases
DUS1, PP3111
UniGene
Hs.514599
Gene Chromosome Location
Chr.17: 82057872-82065821 on Build GRCh38
Transcripts
NM_022156.4 XM_005256393.2 XM_005256394.2 XM_006722288.2 XM_006722289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1303529 c.1096G>A p.E366K Chr.17: 82060020-82060020 on Build GRCh38
4925762 c.1052G>T p.G351V Chr.17: 82060064-82060064 on Build GRCh38
1564124 c.1110G>A p.K370K Chr.17: 82060006-82060006 on Build GRCh38
986020 c.1122G>T p.K374N Chr.17: 82059994-82059994 on Build GRCh38
1178451 c.1090G>A p.G364S Chr.17: 82060026-82060026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372840884 Chr.17: 82060110-82060110 on Build GRCh38
rs200132619 Chr.17: 82060039-82060039 on Build GRCh38
rs1009607568 Chr.17: 82059991-82059993 on Build GRCh38
rs141586689 Chr.17: 82059976-82059976 on Build GRCh38
rs201400745 Chr.17: 82060061-82060061 on Build GRCh38
rs112527177 Chr.17: 82059966-82059966 on Build GRCh38
rs550338959 Chr.17: 82059956-82059956 on Build GRCh38
rs980952219 Chr.17: 82060018-82060018 on Build GRCh38
rs760036782 Chr.17: 82060014-82060014 on Build GRCh38
rs761984352 Chr.17: 82059934-82059934 on Build GRCh38
rs762406276 Chr.17: 82059954-82059954 on Build GRCh38
rs920503975 Chr.17: 82060092-82060092 on Build GRCh38
rs768651289 Chr.17: 82059978-82059978 on Build GRCh38
rs777031875 Chr.17: 82060010-82060010 on Build GRCh38
rs374006789 Chr.17: 82059932-82059932 on Build GRCh38
rs776131195 Chr.17: 82060068-82060068 on Build GRCh38
rs142404033 Chr.17: 82060058-82060058 on Build GRCh38
rs754402667 Chr.17: 82060089-82060090 on Build GRCh38
rs143653555 Chr.17: 82059987-82059987 on Build GRCh38
rs746363915 Chr.17: 82060064-82060064 on Build GRCh38
rs746638846 Chr.17: 82059913-82059913 on Build GRCh38
rs779909564 Chr.17: 82059977-82059977 on Build GRCh38
rs749390254 Chr.17: 82059930-82059930 on Build GRCh38
rs767011917 Chr.17: 82060081-82060081 on Build GRCh38
rs761406134 Chr.17: 82060024-82060024 on Build GRCh38
rs769318809 Chr.17: 82060059-82060059 on Build GRCh38
rs773078135 Chr.17: 82060060-82060060 on Build GRCh38
rs73999366 Chr.17: 82060026-82060026 on Build GRCh38
rs766082361 Chr.17: 82059945-82059946 on Build GRCh38
rs761461277 Chr.17: 82060075-82060075 on Build GRCh38
rs138875382 Chr.17: 82060027-82060027 on Build GRCh38
rs202150137 Chr.17: 82060072-82060074 on Build GRCh38
rs200944932 Chr.17: 82060109-82060109 on Build GRCh38
rs781141688 Chr.17: 82059967-82059967 on Build GRCh38
rs753743821 Chr.17: 82060031-82060031 on Build GRCh38
rs781082570 Chr.17: 82059915-82059915 on Build GRCh38
rs1049467699 Chr.17: 82059894-82059894 on Build GRCh38
rs748284715 Chr.17: 82060055-82060055 on Build GRCh38
rs370491758 Chr.17: 82060066-82060066 on Build GRCh38
rs138475537 Chr.17: 82060049-82060049 on Build GRCh38
rs930463967 Chr.17: 82059892-82059892 on Build GRCh38
rs748372732 Chr.17: 82059983-82059983 on Build GRCh38
rs111336032 Chr.17: 82060022-82060022 on Build GRCh38
rs754347823 Chr.17: 82059952-82059952 on Build GRCh38
rs151063790 Chr.17: 82059993-82059993 on Build GRCh38
rs143848672 Chr.17: 82060082-82060082 on Build GRCh38
rs763523434 Chr.17: 82060021-82060021 on Build GRCh38
rs530491799 Chr.17: 82059960-82059960 on Build GRCh38
rs765207168 Chr.17: 82060111-82060111 on Build GRCh38
rs940604798 Chr.17: 82059929-82059929 on Build GRCh38
rs776993833 Chr.17: 82060062-82060062 on Build GRCh38
rs766852189 Chr.17: 82059948-82059948 on Build GRCh38
rs757912662 Chr.17: 82059899-82059899 on Build GRCh38
rs992405819 Chr.17: 82059999-82059999 on Build GRCh38
rs771292542 Chr.17: 82060001-82060001 on Build GRCh38
rs1025004616 Chr.17: 82060017-82060017 on Build GRCh38
rs751154802 Chr.17: 82060034-82060036 on Build GRCh38
rs202204746 Chr.17: 82059880-82059880 on Build GRCh38
rs745815887 Chr.17: 82059920-82059920 on Build GRCh38
rs756014147 Chr.17: 82060029-82060029 on Build GRCh38
rs779003396 Chr.17: 82059979-82059979 on Build GRCh38
rs766102627 Chr.17: 82060089-82060089 on Build GRCh38
rs373189255 Chr.17: 82060074-82060076 on Build GRCh38
rs779213694 Chr.17: 82059964-82059964 on Build GRCh38
rs367854421 Chr.17: 82059933-82059933 on Build GRCh38
rs750313556 Chr.17: 82059973-82059973 on Build GRCh38
rs780040082 Chr.17: 82059921-82059921 on Build GRCh38
rs772450164 Chr.17: 82059945-82059945 on Build GRCh38
rs777336185 Chr.17: 82059910-82059910 on Build GRCh38
rs779859983 Chr.17: 82060030-82060030 on Build GRCh38
rs532211239 Chr.17: 82059947-82059947 on Build GRCh38
rs960921619 Chr.17: 82060025-82060025 on Build GRCh38
rs577296824 Chr.17: 82060087-82060087 on Build GRCh38
rs767108829 Chr.17: 82060079-82060079 on Build GRCh38

View Full Product Details