Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84154492-84154731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAAF1
Entrez Gene ID
123872
Gene Name
dynein axonemal assembly factor 1
Gene Aliases
CILD13, LRRC50, ODA7
UniGene
Hs.310164
Gene Chromosome Location
Chr.16: 84145260-84178761 on Build GRCh38
Transcripts
NM_178452.5 XM_006721129.2 XM_011522853.2 XM_011522854.2 XM_011522855.2 XM_011522857.2 XM_011522858.2 XM_017022918.1 XM_017022919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4627465 c.503A>G p.N168S Chr.16: 84154727-84154727 on Build GRCh38
4896985 c.432C>T p.I144I Chr.16: 84154656-84154656 on Build GRCh38
1679306 c.460C>T p.R154C Chr.16: 84154684-84154684 on Build GRCh38
974341 c.474G>A p.L158L Chr.16: 84154698-84154698 on Build GRCh38
1380161 c.361C>T p.R121C Chr.16: 84154585-84154585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770583195 Chr.16: 84154686-84154686 on Build GRCh38
rs528511752 Chr.16: 84154558-84154558 on Build GRCh38
rs764728278 Chr.16: 84154705-84154705 on Build GRCh38
rs747051175 Chr.16: 84154571-84154571 on Build GRCh38
rs935602377 Chr.16: 84154582-84154582 on Build GRCh38
rs759955343 Chr.16: 84154620-84154620 on Build GRCh38
rs200946538 Chr.16: 84154685-84154685 on Build GRCh38
rs755250932 Chr.16: 84154544-84154544 on Build GRCh38
rs763360364 Chr.16: 84154695-84154695 on Build GRCh38
rs574238677 Chr.16: 84154684-84154684 on Build GRCh38
rs779780798 Chr.16: 84154663-84154663 on Build GRCh38
rs538582505 Chr.16: 84154618-84154618 on Build GRCh38
rs749246104 Chr.16: 84154596-84154596 on Build GRCh38
rs764149615 Chr.16: 84154623-84154624 on Build GRCh38
rs771510433 Chr.16: 84154682-84154682 on Build GRCh38
rs1027879249 Chr.16: 84154702-84154702 on Build GRCh38
rs534620395 Chr.16: 84154670-84154670 on Build GRCh38
rs374726076 Chr.16: 84154636-84154636 on Build GRCh38
rs147124181 Chr.16: 84154671-84154671 on Build GRCh38
rs765776929 Chr.16: 84154546-84154546 on Build GRCh38
rs114879915 Chr.16: 84154519-84154519 on Build GRCh38
rs371065429 Chr.16: 84154619-84154619 on Build GRCh38
rs376980896 Chr.16: 84154657-84154657 on Build GRCh38
rs757598898 Chr.16: 84154581-84154581 on Build GRCh38
rs749082639 Chr.16: 84154669-84154669 on Build GRCh38
rs368031148 Chr.16: 84154585-84154585 on Build GRCh38
rs143005689 Chr.16: 84154583-84154583 on Build GRCh38
rs199646674 Chr.16: 84154649-84154649 on Build GRCh38
rs953543419 Chr.16: 84154609-84154609 on Build GRCh38
rs764675314 Chr.16: 84154643-84154643 on Build GRCh38
rs373040086 Chr.16: 84154551-84154551 on Build GRCh38
rs138464261 Chr.16: 84154688-84154688 on Build GRCh38
rs762000021 Chr.16: 84154707-84154707 on Build GRCh38
rs569848695 Chr.16: 84154617-84154617 on Build GRCh38
rs148194124 Chr.16: 84154615-84154615 on Build GRCh38
rs765574790 Chr.16: 84154626-84154626 on Build GRCh38
rs768580833 Chr.16: 84154604-84154604 on Build GRCh38
rs201504436 Chr.16: 84154559-84154559 on Build GRCh38
rs751710592 Chr.16: 84154644-84154644 on Build GRCh38
rs775514900 Chr.16: 84154587-84154587 on Build GRCh38
rs34897947 Chr.16: 84154586-84154586 on Build GRCh38
rs113737166 Chr.16: 84154640-84154640 on Build GRCh38
rs773892057 Chr.16: 84154613-84154613 on Build GRCh38
rs932160879 Chr.16: 84154588-84154588 on Build GRCh38
rs753097136 Chr.16: 84154635-84154635 on Build GRCh38
rs559515548 Chr.16: 84154526-84154526 on Build GRCh38
rs752043457 Chr.16: 84154706-84154706 on Build GRCh38
rs200506263 Chr.16: 84154676-84154676 on Build GRCh38
rs752828573 Chr.16: 84154554-84154554 on Build GRCh38
rs148444655 Chr.16: 84154574-84154574 on Build GRCh38
rs756239623 Chr.16: 84154599-84154599 on Build GRCh38
rs143312655 Chr.16: 84154656-84154656 on Build GRCh38

View Full Product Details