Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2840726-2840973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCED1A
Entrez Gene ID
64773
Gene Name
PC-esterase domain containing 1A
Gene Aliases
C20orf81, FAM113A, bA12M19.1
UniGene
Hs.29341
Gene Chromosome Location
Chr.20: 2835314-2841243 on Build GRCh38
Transcripts
NM_001271168.1 XM_005260804.2 NM_022575.3 NM_080413.2

Gene Symbol
VPS16
Entrez Gene ID
64601
Gene Name
VPS16, CORVET/HOPS core subunit
Gene Aliases
hVPS16
UniGene
Hs.269577
Gene Chromosome Location
Chr.20: 2840697-2866732 on Build GRCh38
Transcripts
NM_001271168.1 XM_005260804.2 NM_022575.3 NM_080413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1232365 c.8G>A p.C3Y Chr.20: 2840782-2840782 on Build GRCh38
6850667 c.2T>C p.M1T Chr.20: 2840776-2840776 on Build GRCh38
6350176 c.5A>T p.D2V Chr.20: 2840779-2840779 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745645707 Chr.20: 2840852-2840852 on Build GRCh38
rs2297047 Chr.20: 2840955-2840955 on Build GRCh38
rs759930249 Chr.20: 2840761-2840761 on Build GRCh38
rs199981800 Chr.20: 2840788-2840788 on Build GRCh38
rs557694329 Chr.20: 2840757-2840757 on Build GRCh38
rs749253507 Chr.20: 2840871-2840871 on Build GRCh38
rs1054622477 Chr.20: 2840910-2840910 on Build GRCh38
rs2297046 Chr.20: 2840754-2840754 on Build GRCh38
rs927573465 Chr.20: 2840853-2840853 on Build GRCh38
rs955683745 Chr.20: 2840774-2840774 on Build GRCh38
rs114441777 Chr.20: 2840882-2840882 on Build GRCh38
rs1000523206 Chr.20: 2840764-2840764 on Build GRCh38
rs921442638 Chr.20: 2840885-2840885 on Build GRCh38
rs753261069 Chr.20: 2840771-2840771 on Build GRCh38
rs781537016 Chr.20: 2840841-2840841 on Build GRCh38
rs6076435 Chr.20: 2840833-2840833 on Build GRCh38
rs754204779 Chr.20: 2840759-2840759 on Build GRCh38
rs765728758 Chr.20: 2840768-2840768 on Build GRCh38
rs751133402 Chr.20: 2840813-2840813 on Build GRCh38
rs769783495 Chr.20: 2840855-2840855 on Build GRCh38
rs541974761 Chr.20: 2840914-2840914 on Build GRCh38
rs952947429 Chr.20: 2840909-2840909 on Build GRCh38
rs780130069 Chr.20: 2840866-2840866 on Build GRCh38
rs940004116 Chr.20: 2840954-2840954 on Build GRCh38
rs567061093 Chr.20: 2840769-2840769 on Build GRCh38
rs773252998 Chr.20: 2840745-2840745 on Build GRCh38
rs780757815 Chr.20: 2840850-2840850 on Build GRCh38
rs916902022 Chr.20: 2840755-2840755 on Build GRCh38
rs756959064 Chr.20: 2840835-2840835 on Build GRCh38
rs141415553 Chr.20: 2840792-2840792 on Build GRCh38
rs1023232192 Chr.20: 2840756-2840756 on Build GRCh38
rs557116069 Chr.20: 2840939-2840939 on Build GRCh38

View Full Product Details